Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13918397-13918670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3281334 c.913C>T p.P305S Chr.19: 13918543-13918543 on Build GRCh38
4881382 c.900G>A p.L300L Chr.19: 13918530-13918530 on Build GRCh38
4074717 c.924C>A p.L308L Chr.19: 13918554-13918554 on Build GRCh38
5854783 c.914C>T p.P305L Chr.19: 13918544-13918544 on Build GRCh38
6621095 c.904C>T p.R302W Chr.19: 13918534-13918534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2305776 Chr.19: 13918456-13918456 on Build GRCh38
rs915049706 Chr.19: 13918630-13918630 on Build GRCh38
rs777937050 Chr.19: 13918469-13918469 on Build GRCh38
rs940104278 Chr.19: 13918440-13918440 on Build GRCh38
rs898788135 Chr.19: 13918462-13918462 on Build GRCh38
rs781649512 Chr.19: 13918611-13918611 on Build GRCh38
rs73922801 Chr.19: 13918535-13918535 on Build GRCh38
rs372240909 Chr.19: 13918425-13918425 on Build GRCh38
rs752849813 Chr.19: 13918573-13918573 on Build GRCh38
rs747334868 Chr.19: 13918583-13918583 on Build GRCh38
rs566528989 Chr.19: 13918566-13918566 on Build GRCh38
rs369831348 Chr.19: 13918472-13918472 on Build GRCh38
rs772823974 Chr.19: 13918534-13918534 on Build GRCh38
rs895204373 Chr.19: 13918430-13918430 on Build GRCh38
rs904856105 Chr.19: 13918516-13918516 on Build GRCh38
rs775670544 Chr.19: 13918641-13918641 on Build GRCh38
rs772357873 Chr.19: 13918465-13918465 on Build GRCh38
rs777192749 Chr.19: 13918483-13918483 on Build GRCh38
rs577022787 Chr.19: 13918435-13918435 on Build GRCh38
rs1056582384 Chr.19: 13918426-13918426 on Build GRCh38
rs530533895 Chr.19: 13918540-13918540 on Build GRCh38
rs866968334 Chr.19: 13918564-13918564 on Build GRCh38
rs761333372 Chr.19: 13918506-13918506 on Build GRCh38
rs373362702 Chr.19: 13918488-13918488 on Build GRCh38
rs541500407 Chr.19: 13918532-13918532 on Build GRCh38
rs867466225 Chr.19: 13918512-13918512 on Build GRCh38
rs753863476 Chr.19: 13918546-13918546 on Build GRCh38
rs746324930 Chr.19: 13918619-13918619 on Build GRCh38
rs964409389 Chr.19: 13918552-13918552 on Build GRCh38
rs375465140 Chr.19: 13918623-13918623 on Build GRCh38
rs371171727 Chr.19: 13918574-13918574 on Build GRCh38
rs747416253 Chr.19: 13918471-13918471 on Build GRCh38
rs755081945 Chr.19: 13918551-13918551 on Build GRCh38
rs748365267 Chr.19: 13918463-13918463 on Build GRCh38
rs1025854801 Chr.19: 13918484-13918484 on Build GRCh38
rs747705429 Chr.19: 13918628-13918628 on Build GRCh38
rs1009744427 Chr.19: 13918647-13918647 on Build GRCh38
rs950453818 Chr.19: 13918485-13918487 on Build GRCh38
rs1022114244 Chr.19: 13918649-13918649 on Build GRCh38
rs765356081 Chr.19: 13918570-13918570 on Build GRCh38
rs750222217 Chr.19: 13918563-13918564 on Build GRCh38
rs778117971 Chr.19: 13918575-13918575 on Build GRCh38
rs756317935 Chr.19: 13918608-13918608 on Build GRCh38
rs974180329 Chr.19: 13918446-13918446 on Build GRCh38
rs757661925 Chr.19: 13918598-13918598 on Build GRCh38
rs760717448 Chr.19: 13918432-13918432 on Build GRCh38
rs773875673 Chr.19: 13918492-13918492 on Build GRCh38
rs762420118 Chr.19: 13918485-13918485 on Build GRCh38

View Full Product Details