Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154219783-154220056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf43
Entrez Gene ID
25912
Gene Name
chromosome 1 open reading frame 43
Gene Aliases
HSPC012, NICE-3, NS5ATP4, S863-3
UniGene
Hs.287471
Gene Chromosome Location
Chr.1: 154206701-154220797 on Build GRCh38
Transcripts
NM_001098616.2 NM_001297717.1 NM_001297718.1 NM_001297720.1 NM_001297721.1 NM_001297723.1 NM_015449.3 NM_138740.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466893 c.145C>T p.P49S Chr.1: 154219859-154219859 on Build GRCh38
1334393 c.71T>G p.F24C Chr.1: 154219933-154219933 on Build GRCh38
1583462 c.111C>T p.R37R Chr.1: 154219893-154219893 on Build GRCh38
6281894 c.161C>T p.A54V Chr.1: 154219843-154219843 on Build GRCh38
896531 c.146C>A p.P49H Chr.1: 154219858-154219858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760184444 Chr.1: 154219878-154219878 on Build GRCh38
rs201885596 Chr.1: 154219982-154219983 on Build GRCh38
rs1020939480 Chr.1: 154219997-154219997 on Build GRCh38
rs148815648 Chr.1: 154219923-154219923 on Build GRCh38
rs767965672 Chr.1: 154219846-154219846 on Build GRCh38
rs369133345 Chr.1: 154219868-154219868 on Build GRCh38
rs760432838 Chr.1: 154219916-154219919 on Build GRCh38
rs770345972 Chr.1: 154219949-154219949 on Build GRCh38
rs772739485 Chr.1: 154219817-154219817 on Build GRCh38
rs769318622 Chr.1: 154219816-154219816 on Build GRCh38
rs57040940 Chr.1: 154219997-154219997 on Build GRCh38
rs775927080 Chr.1: 154219981-154219981 on Build GRCh38
rs377453150 Chr.1: 154219877-154219877 on Build GRCh38
rs757270805 Chr.1: 154219864-154219864 on Build GRCh38
rs551231942 Chr.1: 154219987-154219987 on Build GRCh38
rs774711810 Chr.1: 154219825-154219825 on Build GRCh38
rs77669817 Chr.1: 154219973-154219973 on Build GRCh38
rs200230586 Chr.1: 154219998-154219998 on Build GRCh38
rs761953909 Chr.1: 154219977-154219981 on Build GRCh38
rs973474450 Chr.1: 154219990-154219991 on Build GRCh38
rs772611552 Chr.1: 154219935-154219935 on Build GRCh38
rs760988281 Chr.1: 154219982-154219982 on Build GRCh38
rs748531037 Chr.1: 154219987-154219987 on Build GRCh38
rs753893215 Chr.1: 154219862-154219862 on Build GRCh38
rs764430551 Chr.1: 154219984-154219984 on Build GRCh38
rs1040861191 Chr.1: 154219847-154219847 on Build GRCh38
rs748891367 Chr.1: 154219938-154219938 on Build GRCh38
rs764081197 Chr.1: 154219851-154219851 on Build GRCh38
rs771523059 Chr.1: 154219988-154219988 on Build GRCh38
rs943798361 Chr.1: 154219841-154219841 on Build GRCh38
rs200832138 Chr.1: 154219953-154219954 on Build GRCh38
rs766851709 Chr.1: 154219820-154219820 on Build GRCh38
rs397793881 Chr.1: 154219997-154219998 on Build GRCh38
rs754814116 Chr.1: 154219872-154219872 on Build GRCh38
rs985249214 Chr.1: 154220022-154220022 on Build GRCh38
rs34176728 Chr.1: 154219987-154219988 on Build GRCh38
rs781639688 Chr.1: 154219985-154219987 on Build GRCh38
rs1053953663 Chr.1: 154219869-154219869 on Build GRCh38
rs540868389 Chr.1: 154219889-154219889 on Build GRCh38
rs772001814 Chr.1: 154219855-154219863 on Build GRCh38
rs780104619 Chr.1: 154219867-154219867 on Build GRCh38
rs768022779 Chr.1: 154219977-154219977 on Build GRCh38
rs1023864421 Chr.1: 154219931-154219931 on Build GRCh38
rs920695940 Chr.1: 154219991-154219992 on Build GRCh38
rs765408024 Chr.1: 154219977-154219978 on Build GRCh38
rs775455457 Chr.1: 154219865-154219876 on Build GRCh38
rs1040697517 Chr.1: 154219827-154219827 on Build GRCh38
rs761431763 Chr.1: 154219811-154219811 on Build GRCh38
rs199824476 Chr.1: 154219818-154219818 on Build GRCh38
rs760047873 Chr.1: 154219837-154219837 on Build GRCh38
rs752987676 Chr.1: 154219850-154219850 on Build GRCh38
rs755378138 Chr.1: 154219984-154219987 on Build GRCh38
rs1009504915 Chr.1: 154219992-154219992 on Build GRCh38
rs113232145 Chr.1: 154220020-154220020 on Build GRCh38
rs754048800 Chr.1: 154219968-154219969 on Build GRCh38

View Full Product Details