Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26465330-26465575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A1
Entrez Gene ID
11120
Gene Name
butyrophilin subfamily 2 member A1
Gene Aliases
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1
UniGene
Hs.159028
Gene Chromosome Location
Chr.6: 26457904-26476621 on Build GRCh38
Transcripts
NM_001197233.2 NM_001197234.2 NM_007049.4 NM_078476.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2858473 c.890A>T p.K297I Chr.6: 26465404-26465404 on Build GRCh38
3777400 c.844G>A p.E282K Chr.6: 26465358-26465358 on Build GRCh38
7092014 c.886G>A p.E296K Chr.6: 26465358-26465358 on Build GRCh38
6457854 c.818G>A p.R273Q Chr.6: 26465332-26465332 on Build GRCh38
6486372 c.906G>C p.V302V Chr.6: 26465378-26465378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753769800 Chr.6: 26465359-26465359 on Build GRCh38
rs983402401 Chr.6: 26465510-26465510 on Build GRCh38
rs958762616 Chr.6: 26465453-26465453 on Build GRCh38
rs141482323 Chr.6: 26465376-26465376 on Build GRCh38
rs879022557 Chr.6: 26465511-26465511 on Build GRCh38
rs35138509 Chr.6: 26465418-26465419 on Build GRCh38
rs746366684 Chr.6: 26465407-26465407 on Build GRCh38
rs748376681 Chr.6: 26465373-26465373 on Build GRCh38
rs995999029 Chr.6: 26465448-26465448 on Build GRCh38
rs201650489 Chr.6: 26465451-26465451 on Build GRCh38
rs370889094 Chr.6: 26465356-26465356 on Build GRCh38
rs970518801 Chr.6: 26465354-26465354 on Build GRCh38
rs772888822 Chr.6: 26465437-26465437 on Build GRCh38
rs952022160 Chr.6: 26465509-26465509 on Build GRCh38
rs534390301 Chr.6: 26465416-26465416 on Build GRCh38
rs147849789 Chr.6: 26465364-26465364 on Build GRCh38
rs771016574 Chr.6: 26465380-26465380 on Build GRCh38
rs900330174 Chr.6: 26465388-26465388 on Build GRCh38
rs568711992 Chr.6: 26465379-26465379 on Build GRCh38
rs778785394 Chr.6: 26465374-26465374 on Build GRCh38
rs6922353 Chr.6: 26465540-26465540 on Build GRCh38
rs981177558 Chr.6: 26465545-26465545 on Build GRCh38
rs879591713 Chr.6: 26465547-26465550 on Build GRCh38
rs770285409 Chr.6: 26465410-26465410 on Build GRCh38
rs571070905 Chr.6: 26465495-26465495 on Build GRCh38
rs377368650 Chr.6: 26465432-26465432 on Build GRCh38
rs1042075476 Chr.6: 26465357-26465357 on Build GRCh38
rs761576950 Chr.6: 26465424-26465424 on Build GRCh38
rs758556819 Chr.6: 26465524-26465524 on Build GRCh38
rs1027447023 Chr.6: 26465494-26465494 on Build GRCh38
rs750527056 Chr.6: 26465393-26465393 on Build GRCh38

View Full Product Details