Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75087394-75087647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
UniGene
Hs.101302
Gene Chromosome Location
Chr.6: 75084326-75206262 on Build GRCh38
Transcripts
NM_004370.5 NM_080645.2 XM_011535434.1 XM_011535435.1 XM_011535436.2 XM_017010252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875466 c.9153C>A p.S3051R Chr.6: 75087605-75087605 on Build GRCh38
5431011 c.9121C>T p.P3041S Chr.6: 75087637-75087637 on Build GRCh38
1284215 c.9129C>A p.Y3043* Chr.6: 75087629-75087629 on Build GRCh38
4562103 c.9124G>A p.G3042R Chr.6: 75087634-75087634 on Build GRCh38
3875465 c.9157C>G p.P3053A Chr.6: 75087601-75087601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751471471 Chr.6: 75087624-75087624 on Build GRCh38
rs763341868 Chr.6: 75087557-75087557 on Build GRCh38
rs879090705 Chr.6: 75087472-75087472 on Build GRCh38
rs369646700 Chr.6: 75087593-75087593 on Build GRCh38
rs768155096 Chr.6: 75087532-75087532 on Build GRCh38
rs747009147 Chr.6: 75087526-75087526 on Build GRCh38
rs547541301 Chr.6: 75087522-75087522 on Build GRCh38
rs756395971 Chr.6: 75087514-75087514 on Build GRCh38
rs769439797 Chr.6: 75087544-75087544 on Build GRCh38
rs57396313 Chr.6: 75087614-75087614 on Build GRCh38
rs767461073 Chr.6: 75087587-75087587 on Build GRCh38
rs116059284 Chr.6: 75087524-75087524 on Build GRCh38
rs35292916 Chr.6: 75087596-75087596 on Build GRCh38
rs140011165 Chr.6: 75087433-75087433 on Build GRCh38
rs771434132 Chr.6: 75087569-75087569 on Build GRCh38
rs970547 Chr.6: 75087586-75087586 on Build GRCh38
rs867032927 Chr.6: 75087559-75087559 on Build GRCh38
rs267601120 Chr.6: 75087621-75087621 on Build GRCh38
rs752866217 Chr.6: 75087588-75087588 on Build GRCh38
rs565801419 Chr.6: 75087523-75087523 on Build GRCh38
rs765185429 Chr.6: 75087426-75087426 on Build GRCh38
rs1002433115 Chr.6: 75087589-75087589 on Build GRCh38
rs369315573 Chr.6: 75087543-75087543 on Build GRCh38
rs752857440 Chr.6: 75087513-75087513 on Build GRCh38
rs774985994 Chr.6: 75087573-75087573 on Build GRCh38
rs563604000 Chr.6: 75087427-75087427 on Build GRCh38
rs1004806251 Chr.6: 75087449-75087449 on Build GRCh38
rs767762945 Chr.6: 75087508-75087508 on Build GRCh38
rs779895718 Chr.6: 75087615-75087615 on Build GRCh38
rs751344618 Chr.6: 75087592-75087592 on Build GRCh38
rs758519964 Chr.6: 75087572-75087572 on Build GRCh38
rs371725823 Chr.6: 75087550-75087550 on Build GRCh38

View Full Product Details