Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55120346-55120613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20B
Entrez Gene ID
166979
Gene Name
cell division cycle 20B
Gene Aliases
G6VTS76519
UniGene
Hs.669184
Gene Chromosome Location
Chr.5: 55112971-55173177 on Build GRCh38
Transcripts
NM_001145734.2 NM_001170402.1 NM_152623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
364661 c.1216-1G>T p.? Chr.5: 55120551-55120551 on Build GRCh38
6888618 c.1265G>A p.G422E Chr.5: 55120501-55120501 on Build GRCh38
4670636 c.1264G>T p.G422* Chr.5: 55120502-55120502 on Build GRCh38
371935 c.1324C>A p.P442T Chr.5: 55120442-55120442 on Build GRCh38
5403621 c.1231_1232CC>TT p.P411F Chr.5: 55120534-55120535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769189130 Chr.5: 55120429-55120429 on Build GRCh38
rs1024757331 Chr.5: 55120403-55120403 on Build GRCh38
rs755610525 Chr.5: 55120502-55120502 on Build GRCh38
rs746655038 Chr.5: 55120390-55120390 on Build GRCh38
rs138979071 Chr.5: 55120479-55120479 on Build GRCh38
rs766386686 Chr.5: 55120491-55120491 on Build GRCh38
rs142234339 Chr.5: 55120487-55120487 on Build GRCh38
rs779478947 Chr.5: 55120389-55120389 on Build GRCh38
rs532500094 Chr.5: 55120524-55120524 on Build GRCh38
rs201051311 Chr.5: 55120440-55120440 on Build GRCh38
rs773429830 Chr.5: 55120577-55120577 on Build GRCh38
rs768627685 Chr.5: 55120550-55120550 on Build GRCh38
rs147833348 Chr.5: 55120413-55120413 on Build GRCh38
rs749189261 Chr.5: 55120547-55120547 on Build GRCh38
rs970665403 Chr.5: 55120411-55120411 on Build GRCh38
rs374544307 Chr.5: 55120387-55120387 on Build GRCh38
rs531828662 Chr.5: 55120454-55120454 on Build GRCh38
rs374125586 Chr.5: 55120481-55120481 on Build GRCh38
rs774926432 Chr.5: 55120544-55120544 on Build GRCh38
rs761559128 Chr.5: 55120572-55120572 on Build GRCh38
rs769333455 Chr.5: 55120540-55120540 on Build GRCh38
rs767726535 Chr.5: 55120478-55120478 on Build GRCh38
rs745520452 Chr.5: 55120528-55120528 on Build GRCh38
rs775439560 Chr.5: 55120433-55120433 on Build GRCh38
rs758719562 Chr.5: 55120512-55120512 on Build GRCh38
rs768325446 Chr.5: 55120392-55120392 on Build GRCh38
rs779392014 Chr.5: 55120506-55120506 on Build GRCh38
rs745723019 Chr.5: 55120407-55120407 on Build GRCh38
rs761998526 Chr.5: 55120459-55120459 on Build GRCh38
rs749768291 Chr.5: 55120379-55120379 on Build GRCh38
rs866847456 Chr.5: 55120460-55120460 on Build GRCh38
rs188607678 Chr.5: 55120402-55120402 on Build GRCh38
rs1033477518 Chr.5: 55120464-55120464 on Build GRCh38
rs370277291 Chr.5: 55120438-55120438 on Build GRCh38
rs748574072 Chr.5: 55120511-55120511 on Build GRCh38
rs754387691 Chr.5: 55120494-55120494 on Build GRCh38
rs774292084 Chr.5: 55120555-55120555 on Build GRCh38
rs959538554 Chr.5: 55120546-55120546 on Build GRCh38
rs1020733503 Chr.5: 55120520-55120520 on Build GRCh38
rs1002572362 Chr.5: 55120420-55120420 on Build GRCh38
rs145096410 Chr.5: 55120455-55120455 on Build GRCh38
rs770749493 Chr.5: 55120393-55120393 on Build GRCh38
rs547492617 Chr.5: 55120576-55120576 on Build GRCh38
rs267600654 Chr.5: 55120444-55120444 on Build GRCh38

View Full Product Details