Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7177019-7177244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASGR1
Entrez Gene ID
432
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGPR, ASGPR1, CLEC4H1, HL-1
UniGene
Hs.12056
Gene Chromosome Location
Chr.17: 7173431-7179564 on Build GRCh38
Transcripts
NM_001197216.2 NM_001671.4 XM_011523861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605679 c.209T>C p.L70P Chr.17: 7177055-7177055 on Build GRCh38
1679450 c.209T>A p.L70Q Chr.17: 7177055-7177055 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778849332 Chr.17: 7177091-7177091 on Build GRCh38
rs777416579 Chr.17: 7177210-7177210 on Build GRCh38
rs749036522 Chr.17: 7177215-7177215 on Build GRCh38
rs201901672 Chr.17: 7177159-7177159 on Build GRCh38
rs750485682 Chr.17: 7177183-7177183 on Build GRCh38
rs780799811 Chr.17: 7177169-7177170 on Build GRCh38
rs151284398 Chr.17: 7177073-7177073 on Build GRCh38
rs759946011 Chr.17: 7177052-7177052 on Build GRCh38
rs751585208 Chr.17: 7177092-7177092 on Build GRCh38
rs1004252855 Chr.17: 7177134-7177134 on Build GRCh38
rs1016956127 Chr.17: 7177086-7177086 on Build GRCh38
rs997138874 Chr.17: 7177115-7177115 on Build GRCh38
rs761149848 Chr.17: 7177168-7177168 on Build GRCh38
rs773729622 Chr.17: 7177124-7177124 on Build GRCh38
rs754916419 Chr.17: 7177094-7177094 on Build GRCh38
rs757394929 Chr.17: 7177081-7177081 on Build GRCh38
rs745308724 Chr.17: 7177125-7177125 on Build GRCh38
rs777522225 Chr.17: 7177111-7177111 on Build GRCh38
rs770532163 Chr.17: 7177123-7177123 on Build GRCh38
rs781053274 Chr.17: 7177100-7177100 on Build GRCh38
rs774970972 Chr.17: 7177043-7177043 on Build GRCh38
rs748870259 Chr.17: 7177118-7177118 on Build GRCh38
rs754485669 Chr.17: 7177168-7177169 on Build GRCh38
rs746522471 Chr.17: 7177075-7177075 on Build GRCh38
rs200524151 Chr.17: 7177195-7177195 on Build GRCh38
rs2230869 Chr.17: 7177221-7177221 on Build GRCh38
rs867461476 Chr.17: 7177062-7177062 on Build GRCh38
rs772718366 Chr.17: 7177163-7177163 on Build GRCh38
rs768059642 Chr.17: 7177053-7177053 on Build GRCh38
rs756862247 Chr.17: 7177219-7177219 on Build GRCh38
rs753963480 Chr.17: 7177078-7177078 on Build GRCh38
rs551873737 Chr.17: 7177101-7177101 on Build GRCh38
rs767476051 Chr.17: 7177189-7177189 on Build GRCh38
rs778569666 Chr.17: 7177220-7177220 on Build GRCh38
rs868440899 Chr.17: 7177049-7177049 on Build GRCh38
rs761060045 Chr.17: 7177059-7177059 on Build GRCh38
rs758426772 Chr.17: 7177185-7177185 on Build GRCh38
rs886787481 Chr.17: 7177131-7177131 on Build GRCh38
rs1015179927 Chr.17: 7177169-7177169 on Build GRCh38
rs776957193 Chr.17: 7177175-7177175 on Build GRCh38
rs3215126 Chr.17: 7177189-7177190 on Build GRCh38
rs769539653 Chr.17: 7177110-7177110 on Build GRCh38
rs761940338 Chr.17: 7177179-7177179 on Build GRCh38
rs755953428 Chr.17: 7177193-7177193 on Build GRCh38
rs1006022331 Chr.17: 7177042-7177042 on Build GRCh38
rs770317021 Chr.17: 7177039-7177039 on Build GRCh38
rs183814527 Chr.17: 7177170-7177170 on Build GRCh38
rs753114561 Chr.17: 7177058-7177058 on Build GRCh38
rs752600739 Chr.17: 7177190-7177190 on Build GRCh38
rs747759303 Chr.17: 7177184-7177185 on Build GRCh38

View Full Product Details