Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 69938057-69938312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL19A1
Entrez Gene ID
1310
Gene Name
collagen type XIX alpha 1 chain
Gene Aliases
COL9A1L, D6S228E
UniGene
Hs.444842
Gene Chromosome Location
Chr.6: 69866556-70215063 on Build GRCh38
Transcripts
NM_001858.5 XM_011535437.2 XM_011535438.2 XM_017010253.1 XM_017010254.1 XM_017010255.1 XM_017010256.1 XM_017010257.1 XM_017010258.1 XM_017010259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5495837 c.927A>C p.K309N Chr.6: 69938091-69938091 on Build GRCh38
3629273 c.907C>T p.P303S Chr.6: 69938071-69938071 on Build GRCh38
5495838 c.932A>T p.E311V Chr.6: 69938096-69938096 on Build GRCh38
6632706 c.920G>A p.G307E Chr.6: 69938084-69938084 on Build GRCh38
5528928 c.893G>A p.G298E Chr.6: 69938057-69938057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530296124 Chr.6: 69938213-69938213 on Build GRCh38
rs140192576 Chr.6: 69938088-69938088 on Build GRCh38
rs530922397 Chr.6: 69938077-69938077 on Build GRCh38
rs558872553 Chr.6: 69938104-69938104 on Build GRCh38
rs1013178083 Chr.6: 69938122-69938122 on Build GRCh38
rs761741739 Chr.6: 69938141-69938141 on Build GRCh38
rs771748530 Chr.6: 69938120-69938120 on Build GRCh38
rs539082924 Chr.6: 69938249-69938249 on Build GRCh38
rs764953617 Chr.6: 69938143-69938143 on Build GRCh38
rs547157432 Chr.6: 69938273-69938273 on Build GRCh38
rs570323386 Chr.6: 69938237-69938237 on Build GRCh38
rs915337289 Chr.6: 69938280-69938280 on Build GRCh38
rs775017429 Chr.6: 69938121-69938121 on Build GRCh38
rs963423280 Chr.6: 69938113-69938113 on Build GRCh38
rs973171292 Chr.6: 69938131-69938131 on Build GRCh38
rs78276868 Chr.6: 69938288-69938288 on Build GRCh38
rs28648748 Chr.6: 69938094-69938094 on Build GRCh38
rs368097780 Chr.6: 69938101-69938101 on Build GRCh38
rs775910666 Chr.6: 69938083-69938083 on Build GRCh38
rs199945062 Chr.6: 69938099-69938099 on Build GRCh38
rs57916671 Chr.6: 69938195-69938195 on Build GRCh38
rs117424419 Chr.6: 69938137-69938137 on Build GRCh38
rs974535798 Chr.6: 69938252-69938252 on Build GRCh38
rs760686743 Chr.6: 69938205-69938205 on Build GRCh38
rs534636199 Chr.6: 69938229-69938235 on Build GRCh38
rs60011987 Chr.6: 69938219-69938219 on Build GRCh38
rs946829542 Chr.6: 69938286-69938286 on Build GRCh38
rs745563154 Chr.6: 69938106-69938106 on Build GRCh38
rs9717916 Chr.6: 69938105-69938105 on Build GRCh38
rs371747978 Chr.6: 69938200-69938200 on Build GRCh38
rs1022774334 Chr.6: 69938123-69938123 on Build GRCh38
rs760132719 Chr.6: 69938105-69938106 on Build GRCh38
rs199652343 Chr.6: 69938134-69938134 on Build GRCh38
rs773048873 Chr.6: 69938151-69938151 on Build GRCh38
rs768822350 Chr.6: 69938142-69938142 on Build GRCh38

View Full Product Details