Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75314609-75314882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4900326 c.1141G>A p.E381K Chr.6: 75314691-75314691 on Build GRCh38
3875562 c.1000A>G p.K334E Chr.6: 75314832-75314832 on Build GRCh38
3629965 c.1150C>T p.R384* Chr.6: 75314682-75314682 on Build GRCh38
1672572 c.986G>T p.R329M Chr.6: 75314846-75314846 on Build GRCh38
6514175 c.1128C>T p.L376L Chr.6: 75314704-75314704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147080592 Chr.6: 75314681-75314681 on Build GRCh38
rs771543313 Chr.6: 75314785-75314785 on Build GRCh38
rs756290344 Chr.6: 75314767-75314767 on Build GRCh38
rs377467412 Chr.6: 75314638-75314638 on Build GRCh38
rs889571226 Chr.6: 75314830-75314830 on Build GRCh38
rs565134355 Chr.6: 75314849-75314849 on Build GRCh38
rs369160081 Chr.6: 75314642-75314642 on Build GRCh38
rs778985483 Chr.6: 75314694-75314694 on Build GRCh38
rs745729808 Chr.6: 75314696-75314696 on Build GRCh38
rs766905802 Chr.6: 75314740-75314740 on Build GRCh38
rs746478676 Chr.6: 75314791-75314791 on Build GRCh38
rs141211664 Chr.6: 75314729-75314729 on Build GRCh38
rs759714559 Chr.6: 75314746-75314746 on Build GRCh38
rs768376745 Chr.6: 75314641-75314641 on Build GRCh38
rs767624596 Chr.6: 75314750-75314750 on Build GRCh38
rs751662162 Chr.6: 75314680-75314680 on Build GRCh38
rs768162714 Chr.6: 75314715-75314715 on Build GRCh38
rs867513105 Chr.6: 75314853-75314853 on Build GRCh38
rs540380659 Chr.6: 75314835-75314835 on Build GRCh38
rs780178951 Chr.6: 75314788-75314788 on Build GRCh38
rs149447209 Chr.6: 75314786-75314786 on Build GRCh38
rs771408262 Chr.6: 75314837-75314837 on Build GRCh38
rs746559220 Chr.6: 75314706-75314706 on Build GRCh38
rs181966825 Chr.6: 75314716-75314716 on Build GRCh38
rs931076119 Chr.6: 75314762-75314762 on Build GRCh38
rs775441968 Chr.6: 75314858-75314858 on Build GRCh38
rs919711527 Chr.6: 75314745-75314745 on Build GRCh38
rs754574242 Chr.6: 75314803-75314803 on Build GRCh38
rs151315040 Chr.6: 75314653-75314653 on Build GRCh38
rs763403214 Chr.6: 75314739-75314739 on Build GRCh38
rs753025702 Chr.6: 75314760-75314760 on Build GRCh38
rs773581684 Chr.6: 75314733-75314733 on Build GRCh38
rs866728885 Chr.6: 75314667-75314667 on Build GRCh38
rs1025520310 Chr.6: 75314657-75314657 on Build GRCh38
rs765063750 Chr.6: 75314773-75314773 on Build GRCh38
rs750262495 Chr.6: 75314785-75314785 on Build GRCh38
rs758421123 Chr.6: 75314702-75314702 on Build GRCh38
rs55780050 Chr.6: 75314644-75314644 on Build GRCh38
rs61736188 Chr.6: 75314703-75314703 on Build GRCh38
rs753556981 Chr.6: 75314682-75314682 on Build GRCh38
rs141766419 Chr.6: 75314676-75314676 on Build GRCh38
rs770077609 Chr.6: 75314732-75314732 on Build GRCh38
rs769461657 Chr.6: 75314827-75314827 on Build GRCh38
rs377559714 Chr.6: 75314857-75314857 on Build GRCh38
rs765638620 Chr.6: 75314646-75314646 on Build GRCh38
rs780573829 Chr.6: 75314806-75314806 on Build GRCh38
rs774985091 Chr.6: 75314847-75314847 on Build GRCh38
rs202178049 Chr.6: 75314675-75314675 on Build GRCh38
rs745863788 Chr.6: 75314812-75314812 on Build GRCh38
rs867834619 Chr.6: 75314805-75314805 on Build GRCh38
rs774386197 Chr.6: 75314744-75314744 on Build GRCh38
rs960539845 Chr.6: 75314658-75314658 on Build GRCh38
rs757137460 Chr.6: 75314692-75314692 on Build GRCh38
rs777739895 Chr.6: 75314832-75314833 on Build GRCh38

View Full Product Details