Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38729489-38729762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALM
Entrez Gene ID
130589
Gene Name
galactose mutarotase
Gene Aliases
BLOCK25, GLAT, HEL-S-63p, IBD1
UniGene
Hs.435012
Gene Chromosome Location
Chr.2: 38665910-38734767 on Build GRCh38
Transcripts
NM_138801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3731459 c.635-2_635delAGG p.? Chr.2: 38729554-38729556 on Build GRCh38
6054054 c.679C>G p.P227A Chr.2: 38729600-38729600 on Build GRCh38
721033 c.672G>T p.L224L Chr.2: 38729593-38729593 on Build GRCh38
3183910 c.765delT p.C257fs*13 Chr.2: 38729686-38729686 on Build GRCh38
7369153 c.773C>G p.A258G Chr.2: 38729694-38729694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770288133 Chr.2: 38729652-38729652 on Build GRCh38
rs911411022 Chr.2: 38729684-38729684 on Build GRCh38
rs759728547 Chr.2: 38729686-38729686 on Build GRCh38
rs771246616 Chr.2: 38729679-38729679 on Build GRCh38
rs772967156 Chr.2: 38729704-38729714 on Build GRCh38
rs1053757665 Chr.2: 38729703-38729703 on Build GRCh38
rs139320905 Chr.2: 38729694-38729694 on Build GRCh38
rs902557656 Chr.2: 38729662-38729662 on Build GRCh38
rs773651034 Chr.2: 38729671-38729671 on Build GRCh38
rs369381673 Chr.2: 38729617-38729617 on Build GRCh38
rs778733977 Chr.2: 38729588-38729588 on Build GRCh38
rs376778218 Chr.2: 38729543-38729543 on Build GRCh38
rs893060297 Chr.2: 38729708-38729708 on Build GRCh38
rs774667697 Chr.2: 38729682-38729682 on Build GRCh38
rs973061425 Chr.2: 38729609-38729609 on Build GRCh38
rs764696966 Chr.2: 38729633-38729634 on Build GRCh38
rs373416313 Chr.2: 38729723-38729723 on Build GRCh38
rs369978797 Chr.2: 38729719-38729719 on Build GRCh38
rs544238291 Chr.2: 38729548-38729548 on Build GRCh38
rs140688063 Chr.2: 38729596-38729596 on Build GRCh38
rs779485843 Chr.2: 38729620-38729620 on Build GRCh38
rs986085675 Chr.2: 38729674-38729674 on Build GRCh38
rs756065163 Chr.2: 38729560-38729560 on Build GRCh38
rs372545792 Chr.2: 38729542-38729542 on Build GRCh38
rs757065320 Chr.2: 38729587-38729587 on Build GRCh38
rs751731532 Chr.2: 38729520-38729520 on Build GRCh38
rs763330082 Chr.2: 38729675-38729675 on Build GRCh38
rs753693329 Chr.2: 38729584-38729584 on Build GRCh38
rs771730357 Chr.2: 38729606-38729606 on Build GRCh38
rs1041172332 Chr.2: 38729698-38729698 on Build GRCh38
rs766959943 Chr.2: 38729568-38729568 on Build GRCh38
rs374958454 Chr.2: 38729638-38729638 on Build GRCh38
rs761381275 Chr.2: 38729615-38729616 on Build GRCh38
rs760743319 Chr.2: 38729714-38729714 on Build GRCh38
rs764144026 Chr.2: 38729721-38729721 on Build GRCh38

View Full Product Details