Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35562646-35562907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM166B
Entrez Gene ID
730112
Gene Name
family with sequence similarity 166 member B
Gene Aliases
-
UniGene
Hs.98959
Gene Chromosome Location
Chr.9: 35561830-35563899 on Build GRCh38
Transcripts
NM_001099951.3 NM_001164310.2 NM_001287238.1 NM_001287239.1 XM_011518028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027064 c.532T>G p.F178V Chr.9: 35562661-35562661 on Build GRCh38
6658838 c.471G>A p.P157P Chr.9: 35562896-35562896 on Build GRCh38
6658839 c.471G>A p.P157P Chr.9: 35562896-35562896 on Build GRCh38
5027065 c.532T>G p.F178V Chr.9: 35562661-35562661 on Build GRCh38
6295282 c.507C>A p.S169S Chr.9: 35562686-35562686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755740475 Chr.9: 35562725-35562728 on Build GRCh38
rs778950792 Chr.9: 35562813-35562813 on Build GRCh38
rs770916723 Chr.9: 35562868-35562868 on Build GRCh38
rs774546651 Chr.9: 35562712-35562712 on Build GRCh38
rs780249704 Chr.9: 35562834-35562834 on Build GRCh38
rs747044569 Chr.9: 35562747-35562747 on Build GRCh38
rs542699567 Chr.9: 35562823-35562823 on Build GRCh38
rs932484411 Chr.9: 35562827-35562827 on Build GRCh38
rs962714882 Chr.9: 35562761-35562761 on Build GRCh38
rs368744999 Chr.9: 35562673-35562673 on Build GRCh38
rs543478564 Chr.9: 35562763-35562763 on Build GRCh38
rs770167550 Chr.9: 35562865-35562865 on Build GRCh38
rs775523774 Chr.9: 35562726-35562726 on Build GRCh38
rs762289527 Chr.9: 35562736-35562736 on Build GRCh38
rs750098374 Chr.9: 35562709-35562710 on Build GRCh38
rs1004932896 Chr.9: 35562764-35562764 on Build GRCh38
rs777416693 Chr.9: 35562805-35562805 on Build GRCh38
rs199821364 Chr.9: 35562731-35562731 on Build GRCh38
rs771729993 Chr.9: 35562705-35562706 on Build GRCh38
rs747860317 Chr.9: 35562685-35562685 on Build GRCh38
rs573830796 Chr.9: 35562803-35562803 on Build GRCh38
rs768975911 Chr.9: 35562838-35562838 on Build GRCh38
rs753569138 Chr.9: 35562802-35562803 on Build GRCh38
rs780785364 Chr.9: 35562679-35562679 on Build GRCh38
rs754595181 Chr.9: 35562678-35562678 on Build GRCh38
rs373211462 Chr.9: 35562713-35562713 on Build GRCh38
rs766946841 Chr.9: 35562698-35562699 on Build GRCh38
rs776830848 Chr.9: 35562853-35562853 on Build GRCh38
rs750960160 Chr.9: 35562750-35562750 on Build GRCh38
rs763454320 Chr.9: 35562870-35562870 on Build GRCh38
rs995879392 Chr.9: 35562713-35562713 on Build GRCh38
rs749176744 Chr.9: 35562700-35562700 on Build GRCh38
rs757226823 Chr.9: 35562812-35562812 on Build GRCh38
rs765773858 Chr.9: 35562749-35562749 on Build GRCh38
rs777626724 Chr.9: 35562696-35562696 on Build GRCh38
rs770237079 Chr.9: 35562743-35562743 on Build GRCh38
rs376934439 Chr.9: 35562714-35562714 on Build GRCh38
rs752223659 Chr.9: 35562804-35562804 on Build GRCh38
rs754998499 Chr.9: 35562827-35562828 on Build GRCh38
rs973837666 Chr.9: 35562802-35562802 on Build GRCh38
rs1015179863 Chr.9: 35562773-35562773 on Build GRCh38
rs765864465 Chr.9: 35562669-35562669 on Build GRCh38
rs374141690 Chr.9: 35562854-35562854 on Build GRCh38
rs772215011 Chr.9: 35562831-35562831 on Build GRCh38
rs771049897 Chr.9: 35562705-35562705 on Build GRCh38
rs766200297 Chr.9: 35562801-35562802 on Build GRCh38
rs760959531 Chr.9: 35562727-35562727 on Build GRCh38
rs371526938 Chr.9: 35562836-35562836 on Build GRCh38
rs556974950 Chr.9: 35562770-35562770 on Build GRCh38
rs758999877 Chr.9: 35562801-35562801 on Build GRCh38
rs778726661 Chr.9: 35562885-35562886 on Build GRCh38
rs755804958 Chr.9: 35562691-35562691 on Build GRCh38
rs761243421 Chr.9: 35562675-35562676 on Build GRCh38

View Full Product Details