Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27759443-27759704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76A
Entrez Gene ID
199870
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
UniGene
Hs.469359
Gene Chromosome Location
Chr.1: 27725979-27762915 on Build GRCh38
Transcripts
NM_001143912.1 NM_001143913.1 NM_001143914.1 NM_001143915.1 NM_152660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3789843 c.894G>A p.M298I Chr.1: 27759582-27759582 on Build GRCh38
5956701 c.751G>T p.A251S Chr.1: 27759541-27759541 on Build GRCh38
6302339 c.838-1G>T p.? Chr.1: 27759525-27759525 on Build GRCh38
7292106 c.831A>G p.Q277Q Chr.1: 27759621-27759621 on Build GRCh38
3487726 c.773C>T p.S258L Chr.1: 27759563-27759563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747668325 Chr.1: 27759573-27759573 on Build GRCh38
rs202236285 Chr.1: 27759558-27759558 on Build GRCh38
rs766674517 Chr.1: 27759480-27759480 on Build GRCh38
rs767651186 Chr.1: 27759611-27759611 on Build GRCh38
rs758362731 Chr.1: 27759655-27759655 on Build GRCh38
rs375363374 Chr.1: 27759496-27759496 on Build GRCh38
rs767597454 Chr.1: 27759488-27759488 on Build GRCh38
rs980345071 Chr.1: 27759645-27759645 on Build GRCh38
rs904581997 Chr.1: 27759537-27759537 on Build GRCh38
rs752340683 Chr.1: 27759662-27759662 on Build GRCh38
rs778456969 Chr.1: 27759646-27759646 on Build GRCh38
rs750411172 Chr.1: 27759647-27759647 on Build GRCh38
rs758231941 Chr.1: 27759535-27759535 on Build GRCh38
rs746630944 Chr.1: 27759546-27759546 on Build GRCh38
rs895743783 Chr.1: 27759471-27759472 on Build GRCh38
rs759740785 Chr.1: 27759599-27759599 on Build GRCh38
rs763384928 Chr.1: 27759589-27759589 on Build GRCh38
rs188215445 Chr.1: 27759487-27759487 on Build GRCh38
rs778879776 Chr.1: 27759512-27759512 on Build GRCh38
rs754732403 Chr.1: 27759612-27759612 on Build GRCh38
rs745658829 Chr.1: 27759530-27759530 on Build GRCh38
rs780695065 Chr.1: 27759565-27759565 on Build GRCh38
rs779660616 Chr.1: 27759540-27759540 on Build GRCh38
rs770416312 Chr.1: 27759574-27759574 on Build GRCh38
rs570314153 Chr.1: 27759615-27759615 on Build GRCh38
rs774613464 Chr.1: 27759594-27759594 on Build GRCh38
rs757164138 Chr.1: 27759500-27759500 on Build GRCh38
rs751875896 Chr.1: 27759486-27759486 on Build GRCh38
rs773892912 Chr.1: 27759583-27759583 on Build GRCh38
rs145344355 Chr.1: 27759628-27759628 on Build GRCh38
rs968800535 Chr.1: 27759639-27759639 on Build GRCh38
rs369422673 Chr.1: 27759553-27759553 on Build GRCh38

View Full Product Details