Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22601917-22602154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
-
UniGene
Hs.743508
Gene Chromosome Location
Chr.8: 22599601-22604150 on Build GRCh38
Transcripts
NM_001013842.2 NM_001198827.1 NM_173686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616736 c.676T>C p.S226P Chr.8: 22601990-22601990 on Build GRCh38
5409216 c.688C>T p.P230S Chr.8: 22602002-22602002 on Build GRCh38
3898960 c.663C>T p.P221P Chr.8: 22601977-22601977 on Build GRCh38
7358751 c.701C>T p.P234L Chr.8: 22602015-22602015 on Build GRCh38
4883684 c.712C>A p.P238T Chr.8: 22602026-22602026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954579202 Chr.8: 22601958-22601958 on Build GRCh38
rs772940202 Chr.8: 22601977-22601977 on Build GRCh38
rs767010178 Chr.8: 22602091-22602091 on Build GRCh38
rs73549908 Chr.8: 22601980-22601980 on Build GRCh38
rs764378036 Chr.8: 22601936-22601936 on Build GRCh38
rs865908058 Chr.8: 22602013-22602013 on Build GRCh38
rs375402426 Chr.8: 22602021-22602021 on Build GRCh38
rs753817329 Chr.8: 22602112-22602112 on Build GRCh38
rs73672798 Chr.8: 22602016-22602016 on Build GRCh38
rs777214555 Chr.8: 22602045-22602045 on Build GRCh38
rs552783570 Chr.8: 22601978-22601978 on Build GRCh38
rs752247023 Chr.8: 22602093-22602093 on Build GRCh38
rs750074742 Chr.8: 22601944-22601944 on Build GRCh38
rs765643934 Chr.8: 22602053-22602053 on Build GRCh38
rs761231392 Chr.8: 22602018-22602018 on Build GRCh38
rs374972810 Chr.8: 22601952-22601952 on Build GRCh38
rs753648927 Chr.8: 22601969-22601969 on Build GRCh38
rs777530143 Chr.8: 22602104-22602104 on Build GRCh38
rs751158843 Chr.8: 22602069-22602069 on Build GRCh38
rs768152131 Chr.8: 22602055-22602057 on Build GRCh38
rs748066188 Chr.8: 22602000-22602000 on Build GRCh38
rs759245401 Chr.8: 22602077-22602077 on Build GRCh38
rs150335262 Chr.8: 22601961-22601961 on Build GRCh38
rs756058209 Chr.8: 22602094-22602094 on Build GRCh38
rs754421055 Chr.8: 22601997-22601997 on Build GRCh38
rs762046672 Chr.8: 22602050-22602050 on Build GRCh38
rs941179808 Chr.8: 22601967-22601967 on Build GRCh38
rs149466621 Chr.8: 22601991-22601991 on Build GRCh38
rs777593871 Chr.8: 22602008-22602008 on Build GRCh38
rs748084854 Chr.8: 22601970-22601970 on Build GRCh38
rs780697538 Chr.8: 22601999-22601999 on Build GRCh38
rs758051155 Chr.8: 22601945-22601945 on Build GRCh38
rs756075181 Chr.8: 22602004-22602004 on Build GRCh38
rs781069791 Chr.8: 22601966-22601966 on Build GRCh38
rs760660211 Chr.8: 22601940-22601940 on Build GRCh38
rs867186924 Chr.8: 22602064-22602064 on Build GRCh38
rs1016385124 Chr.8: 22601946-22601946 on Build GRCh38
rs147403979 Chr.8: 22601975-22601975 on Build GRCh38
rs774648856 Chr.8: 22602012-22602012 on Build GRCh38
rs761319510 Chr.8: 22601987-22601987 on Build GRCh38
rs368018226 Chr.8: 22602035-22602035 on Build GRCh38
rs147143680 Chr.8: 22602009-22602009 on Build GRCh38
rs776080445 Chr.8: 22601984-22601984 on Build GRCh38
rs1030190573 Chr.8: 22601956-22601956 on Build GRCh38
rs917745239 Chr.8: 22602032-22602032 on Build GRCh38
rs367926851 Chr.8: 22601955-22601955 on Build GRCh38
rs778909516 Chr.8: 22602117-22602117 on Build GRCh38
rs375493498 Chr.8: 22601996-22601996 on Build GRCh38
rs866861080 Chr.8: 22602002-22602002 on Build GRCh38
rs139447220 Chr.8: 22602015-22602015 on Build GRCh38
rs764512329 Chr.8: 22601988-22601988 on Build GRCh38
rs756943962 Chr.8: 22602116-22602116 on Build GRCh38
rs145113524 Chr.8: 22601995-22601995 on Build GRCh38
rs371627153 Chr.8: 22602010-22602010 on Build GRCh38

View Full Product Details