Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136994863-136995108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC3
Entrez Gene ID
9022
Gene Name
chloride intracellular channel 3
Gene Aliases
-
UniGene
Hs.64746
Gene Chromosome Location
Chr.9: 136994608-136997929 on Build GRCh38
Transcripts
NM_004669.2 XM_017015281.1 XM_017015282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408809 c.457G>T p.E153* Chr.9: 136995025-136995025 on Build GRCh38
7447743 c.499G>A p.D167N Chr.9: 136994983-136994983 on Build GRCh38
6412662 c.427C>A p.R143S Chr.9: 136995055-136995055 on Build GRCh38
5875908 c.381delG p.Y128fs*>109 Chr.9: 136995101-136995101 on Build GRCh38
6411014 c.446A>G p.E149G Chr.9: 136995036-136995036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759869881 Chr.9: 136994910-136994910 on Build GRCh38
rs11145966 Chr.9: 136994936-136994936 on Build GRCh38
rs960748605 Chr.9: 136995045-136995045 on Build GRCh38
rs764518381 Chr.9: 136994977-136994977 on Build GRCh38
rs111505970 Chr.9: 136994957-136994957 on Build GRCh38
rs932903474 Chr.9: 136994894-136994894 on Build GRCh38
rs776802844 Chr.9: 136994882-136994882 on Build GRCh38
rs890124701 Chr.9: 136994927-136994927 on Build GRCh38
rs757765288 Chr.9: 136995015-136995015 on Build GRCh38
rs755773085 Chr.9: 136995057-136995057 on Build GRCh38
rs980713259 Chr.9: 136995002-136995002 on Build GRCh38
rs766962392 Chr.9: 136995020-136995020 on Build GRCh38
rs988075339 Chr.9: 136995076-136995076 on Build GRCh38
rs371960731 Chr.9: 136995000-136995000 on Build GRCh38
rs900116073 Chr.9: 136994888-136994888 on Build GRCh38
rs943378832 Chr.9: 136995038-136995038 on Build GRCh38
rs919118849 Chr.9: 136994969-136994969 on Build GRCh38
rs946897574 Chr.9: 136994881-136994881 on Build GRCh38
rs969745559 Chr.9: 136995001-136995001 on Build GRCh38
rs374553474 Chr.9: 136994944-136994944 on Build GRCh38
rs11145965 Chr.9: 136994933-136994933 on Build GRCh38
rs749865066 Chr.9: 136995037-136995037 on Build GRCh38
rs780016514 Chr.9: 136994903-136994904 on Build GRCh38
rs779736714 Chr.9: 136995072-136995072 on Build GRCh38
rs1039052232 Chr.9: 136995053-136995053 on Build GRCh38
rs981585134 Chr.9: 136994968-136994968 on Build GRCh38
rs929116679 Chr.9: 136994998-136994998 on Build GRCh38
rs897554800 Chr.9: 136994963-136994964 on Build GRCh38
rs575936365 Chr.9: 136994939-136994939 on Build GRCh38
rs763287268 Chr.9: 136994967-136994967 on Build GRCh38
rs749193286 Chr.9: 136995024-136995025 on Build GRCh38

View Full Product Details