Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93201831-93202104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
UniGene
Hs.482873
Gene Chromosome Location
Chr.1: 93179802-93279037 on Build GRCh38
Transcripts
NM_001306076.1 NM_206886.4 XM_011541361.2 XM_011541372.2 XM_017001154.1 XM_017001155.1 XM_017001156.1 XM_017001157.1 XM_017001158.1 XM_017001159.1 XM_017001160.1 XM_017001161.1 XM_017001162.1 XM_017001163.1 XM_017001164.1 XM_017001165.1 XM_017001166.1 XM_017001167.1 XM_017001168.1 XM_017001169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913378 c.1060C>T p.R354* Chr.1: 93201899-93201899 on Build GRCh38
252971 c.1120delA p.V377fs*15 Chr.1: 93201959-93201959 on Build GRCh38
913379 c.1115A>C p.Q372P Chr.1: 93201954-93201954 on Build GRCh38
6364885 c.1120A>T p.K374* Chr.1: 93201959-93201959 on Build GRCh38
6294755 c.1095G>T p.E365D Chr.1: 93201934-93201934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757200472 Chr.1: 93201939-93201939 on Build GRCh38
rs765889131 Chr.1: 93201904-93201904 on Build GRCh38
rs771488979 Chr.1: 93201862-93201863 on Build GRCh38
rs764931902 Chr.1: 93201919-93201919 on Build GRCh38
rs559419360 Chr.1: 93201929-93201929 on Build GRCh38
rs549421415 Chr.1: 93202023-93202023 on Build GRCh38
rs768816831 Chr.1: 93201955-93201955 on Build GRCh38
rs774644240 Chr.1: 93201862-93201862 on Build GRCh38
rs746565576 Chr.1: 93201958-93201959 on Build GRCh38
rs754657117 Chr.1: 93201914-93201914 on Build GRCh38
rs777667910 Chr.1: 93201928-93201928 on Build GRCh38
rs200045120 Chr.1: 93201899-93201899 on Build GRCh38
rs752487584 Chr.1: 93201921-93201921 on Build GRCh38
rs982599544 Chr.1: 93201916-93201916 on Build GRCh38
rs778672296 Chr.1: 93202011-93202011 on Build GRCh38
rs774609342 Chr.1: 93201956-93201956 on Build GRCh38
rs527899280 Chr.1: 93201900-93201900 on Build GRCh38
rs375147622 Chr.1: 93201910-93201910 on Build GRCh38
rs772448332 Chr.1: 93201966-93201966 on Build GRCh38
rs941219797 Chr.1: 93201970-93201970 on Build GRCh38
rs762223740 Chr.1: 93201872-93201872 on Build GRCh38
rs779262598 Chr.1: 93201959-93201959 on Build GRCh38
rs767958287 Chr.1: 93201882-93201882 on Build GRCh38
rs748500228 Chr.1: 93201959-93201959 on Build GRCh38
rs780283117 Chr.1: 93201942-93201942 on Build GRCh38
rs749381469 Chr.1: 93201949-93201949 on Build GRCh38
rs758184693 Chr.1: 93201927-93201927 on Build GRCh38

View Full Product Details