Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85582491-85582760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYR61
Entrez Gene ID
3491
Gene Name
cysteine rich angiogenic inducer 61
Gene Aliases
CCN1, GIG1, IGFBP10
UniGene
Hs.8867
Gene Chromosome Location
Chr.1: 85580761-85583967 on Build GRCh38
Transcripts
NM_001554.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348936 c.760G>T p.D254Y Chr.1: 85582541-85582541 on Build GRCh38
6498127 c.726C>T p.T242T Chr.1: 85582507-85582507 on Build GRCh38
3977935 c.717C>T p.C239C Chr.1: 85582498-85582498 on Build GRCh38
3806006 c.769G>C p.E257Q Chr.1: 85582550-85582550 on Build GRCh38
4854614 c.796A>T p.I266F Chr.1: 85582577-85582577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777112490 Chr.1: 85582562-85582562 on Build GRCh38
rs777361186 Chr.1: 85582645-85582645 on Build GRCh38
rs752431884 Chr.1: 85582642-85582642 on Build GRCh38
rs369974800 Chr.1: 85582644-85582644 on Build GRCh38
rs919894583 Chr.1: 85582539-85582539 on Build GRCh38
rs1055615804 Chr.1: 85582640-85582640 on Build GRCh38
rs75947336 Chr.1: 85582584-85582584 on Build GRCh38
rs777365838 Chr.1: 85582668-85582669 on Build GRCh38
rs755938076 Chr.1: 85582527-85582527 on Build GRCh38
rs770173391 Chr.1: 85582662-85582662 on Build GRCh38
rs746492611 Chr.1: 85582647-85582647 on Build GRCh38
rs749074854 Chr.1: 85582529-85582529 on Build GRCh38
rs766218988 Chr.1: 85582610-85582610 on Build GRCh38
rs771284240 Chr.1: 85582561-85582561 on Build GRCh38
rs754796443 Chr.1: 85582622-85582622 on Build GRCh38
rs770045627 Chr.1: 85582588-85582588 on Build GRCh38
rs1046942147 Chr.1: 85582558-85582558 on Build GRCh38
rs779908967 Chr.1: 85582528-85582528 on Build GRCh38
rs373000437 Chr.1: 85582593-85582593 on Build GRCh38
rs1003234344 Chr.1: 85582639-85582639 on Build GRCh38
rs765129171 Chr.1: 85582727-85582727 on Build GRCh38
rs377358947 Chr.1: 85582682-85582682 on Build GRCh38
rs761766701 Chr.1: 85582675-85582675 on Build GRCh38
rs189866774 Chr.1: 85582672-85582672 on Build GRCh38
rs772560592 Chr.1: 85582556-85582556 on Build GRCh38
rs778674730 Chr.1: 85582536-85582536 on Build GRCh38
rs200203130 Chr.1: 85582535-85582535 on Build GRCh38
rs111410670 Chr.1: 85582739-85582739 on Build GRCh38
rs753851213 Chr.1: 85582719-85582719 on Build GRCh38
rs748958257 Chr.1: 85582723-85582723 on Build GRCh38
rs112232895 Chr.1: 85582737-85582737 on Build GRCh38
rs377332597 Chr.1: 85582723-85582723 on Build GRCh38
rs139284008 Chr.1: 85582524-85582524 on Build GRCh38
rs909867388 Chr.1: 85582591-85582591 on Build GRCh38
rs775761908 Chr.1: 85582663-85582663 on Build GRCh38
rs1011321526 Chr.1: 85582650-85582650 on Build GRCh38
rs759891344 Chr.1: 85582573-85582573 on Build GRCh38
rs763172280 Chr.1: 85582669-85582669 on Build GRCh38
rs769402244 Chr.1: 85582534-85582535 on Build GRCh38
rs922667068 Chr.1: 85582517-85582517 on Build GRCh38
rs955636453 Chr.1: 85582713-85582713 on Build GRCh38
rs764029932 Chr.1: 85582520-85582520 on Build GRCh38
rs368750850 Chr.1: 85582590-85582590 on Build GRCh38
rs775710634 Chr.1: 85582589-85582589 on Build GRCh38
rs373437549 Chr.1: 85582652-85582652 on Build GRCh38
rs778805266 Chr.1: 85582623-85582623 on Build GRCh38
rs755450244 Chr.1: 85582708-85582708 on Build GRCh38
rs746322786 Chr.1: 85582653-85582653 on Build GRCh38
rs561300948 Chr.1: 85582611-85582611 on Build GRCh38
rs376931984 Chr.1: 85582541-85582541 on Build GRCh38
rs747560642 Chr.1: 85582557-85582557 on Build GRCh38

View Full Product Details