Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45455841-45456066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_030582.3 NM_130444.2 NM_130445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830171 c.438G>A p.E146E Chr.21: 45455968-45455968 on Build GRCh38
7271659 c.353C>T p.A118V Chr.21: 45455883-45455883 on Build GRCh38
5767870 c.373G>C p.V125L Chr.21: 45455903-45455903 on Build GRCh38
6526323 c.339G>A p.E113E Chr.21: 45455869-45455869 on Build GRCh38
6533228 c.405G>C p.E135D Chr.21: 45455935-45455935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573521157 Chr.21: 45455983-45455983 on Build GRCh38
rs750868738 Chr.21: 45455867-45455867 on Build GRCh38
rs114139997 Chr.21: 45455861-45455861 on Build GRCh38
rs752318886 Chr.21: 45455947-45455947 on Build GRCh38
rs61759533 Chr.21: 45455960-45455960 on Build GRCh38
rs756742580 Chr.21: 45456027-45456027 on Build GRCh38
rs200284308 Chr.21: 45456045-45456045 on Build GRCh38
rs766977257 Chr.21: 45456009-45456009 on Build GRCh38
rs979909932 Chr.21: 45455910-45455910 on Build GRCh38
rs777996655 Chr.21: 45455893-45455893 on Build GRCh38
rs999764376 Chr.21: 45456030-45456030 on Build GRCh38
rs745610940 Chr.21: 45455895-45455895 on Build GRCh38
rs761130894 Chr.21: 45455994-45455994 on Build GRCh38
rs766711520 Chr.21: 45455997-45455997 on Build GRCh38
rs368269918 Chr.21: 45455928-45455928 on Build GRCh38
rs201773944 Chr.21: 45456032-45456032 on Build GRCh38
rs557923229 Chr.21: 45455879-45455879 on Build GRCh38
rs769054779 Chr.21: 45455981-45455981 on Build GRCh38
rs781126594 Chr.21: 45455974-45455974 on Build GRCh38
rs762591091 Chr.21: 45456002-45456002 on Build GRCh38
rs768823718 Chr.21: 45455911-45455911 on Build GRCh38
rs758099059 Chr.21: 45455949-45455949 on Build GRCh38
rs761528899 Chr.21: 45455925-45455925 on Build GRCh38
rs983804503 Chr.21: 45455937-45455937 on Build GRCh38
rs780515185 Chr.21: 45456000-45456000 on Build GRCh38
rs780835386 Chr.21: 45455894-45455894 on Build GRCh38
rs767745134 Chr.21: 45455866-45455866 on Build GRCh38
rs762210408 Chr.21: 45455920-45455920 on Build GRCh38
rs748336591 Chr.21: 45456048-45456048 on Build GRCh38
rs766966736 Chr.21: 45456029-45456029 on Build GRCh38
rs780033569 Chr.21: 45455980-45455980 on Build GRCh38
rs757851697 Chr.21: 45455883-45455883 on Build GRCh38
rs773715846 Chr.21: 45455993-45455993 on Build GRCh38
rs777309478 Chr.21: 45455884-45455884 on Build GRCh38
rs570051194 Chr.21: 45455904-45455904 on Build GRCh38
rs570643977 Chr.21: 45455950-45455950 on Build GRCh38
rs768133727 Chr.21: 45455922-45455922 on Build GRCh38
rs748188616 Chr.21: 45455984-45455984 on Build GRCh38
rs756054515 Chr.21: 45456043-45456043 on Build GRCh38
rs771299625 Chr.21: 45456009-45456010 on Build GRCh38
rs372080967 Chr.21: 45455902-45455902 on Build GRCh38
rs62000459 Chr.21: 45455878-45455878 on Build GRCh38
rs770965772 Chr.21: 45455889-45455889 on Build GRCh38
rs766689573 Chr.21: 45455935-45455935 on Build GRCh38
rs200559406 Chr.21: 45455903-45455903 on Build GRCh38
rs947330907 Chr.21: 45455989-45455989 on Build GRCh38
rs761038457 Chr.21: 45455930-45455930 on Build GRCh38
rs537323761 Chr.21: 45455913-45455913 on Build GRCh38
rs763670793 Chr.21: 45456013-45456013 on Build GRCh38
rs368985827 Chr.21: 45456044-45456044 on Build GRCh38
rs374634058 Chr.21: 45455860-45455860 on Build GRCh38
rs745814977 Chr.21: 45455975-45455975 on Build GRCh38
rs751138469 Chr.21: 45455959-45455959 on Build GRCh38
rs62000960 Chr.21: 45456047-45456047 on Build GRCh38
rs754577294 Chr.21: 45455875-45455875 on Build GRCh38
rs777041346 Chr.21: 45455999-45455999 on Build GRCh38
rs751226499 Chr.21: 45456017-45456017 on Build GRCh38
rs746915433 Chr.21: 45455888-45455888 on Build GRCh38

View Full Product Details