Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7023980-7024205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6B
Entrez Gene ID
255877
Gene Name
B-cell CLL/lymphoma 6B
Gene Aliases
BAZF, ZBTB28, ZNF62
UniGene
Hs.22575
Gene Chromosome Location
Chr.17: 7023050-7029642 on Build GRCh38
Transcripts
NM_181844.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069118 c.267C>T p.F89F Chr.17: 7024170-7024170 on Build GRCh38
2976034 c.253G>A p.E85K Chr.17: 7024156-7024156 on Build GRCh38
4069120 c.268G>A p.A90T Chr.17: 7024171-7024171 on Build GRCh38
4069114 c.252C>T p.P84P Chr.17: 7024155-7024155 on Build GRCh38
6610762 c.202C>T p.R68W Chr.17: 7024105-7024105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033358477 Chr.17: 7024004-7024004 on Build GRCh38
rs766293348 Chr.17: 7024060-7024060 on Build GRCh38
rs372529088 Chr.17: 7024063-7024063 on Build GRCh38
rs1002845184 Chr.17: 7024145-7024145 on Build GRCh38
rs200688640 Chr.17: 7024112-7024112 on Build GRCh38
rs894391596 Chr.17: 7024178-7024178 on Build GRCh38
rs375084496 Chr.17: 7024072-7024072 on Build GRCh38
rs757738546 Chr.17: 7024092-7024092 on Build GRCh38
rs312461 Chr.17: 7024031-7024031 on Build GRCh38
rs762779920 Chr.17: 7024039-7024039 on Build GRCh38
rs202178963 Chr.17: 7024131-7024131 on Build GRCh38
rs756736408 Chr.17: 7024171-7024171 on Build GRCh38
rs971255128 Chr.17: 7024053-7024053 on Build GRCh38
rs754759982 Chr.17: 7024064-7024064 on Build GRCh38
rs772415626 Chr.17: 7024105-7024105 on Build GRCh38
rs779412938 Chr.17: 7024094-7024094 on Build GRCh38
rs977660436 Chr.17: 7024179-7024179 on Build GRCh38
rs747201143 Chr.17: 7024106-7024106 on Build GRCh38
rs773241303 Chr.17: 7024032-7024032 on Build GRCh38
rs372366130 Chr.17: 7024170-7024170 on Build GRCh38
rs754062790 Chr.17: 7024147-7024147 on Build GRCh38
rs990103242 Chr.17: 7024152-7024152 on Build GRCh38
rs551770853 Chr.17: 7024029-7024029 on Build GRCh38
rs920846763 Chr.17: 7024096-7024096 on Build GRCh38
rs752390604 Chr.17: 7024069-7024069 on Build GRCh38
rs1000145965 Chr.17: 7024003-7024003 on Build GRCh38
rs763759900 Chr.17: 7024167-7024167 on Build GRCh38
rs530841780 Chr.17: 7024067-7024067 on Build GRCh38
rs967203876 Chr.17: 7024161-7024161 on Build GRCh38
rs1034377521 Chr.17: 7024146-7024146 on Build GRCh38
rs779287864 Chr.17: 7024070-7024071 on Build GRCh38
rs748779304 Chr.17: 7024089-7024089 on Build GRCh38
rs1013169816 Chr.17: 7024030-7024030 on Build GRCh38
rs767386551 Chr.17: 7024149-7024149 on Build GRCh38
rs374882649 Chr.17: 7024132-7024132 on Build GRCh38
rs898699839 Chr.17: 7024076-7024076 on Build GRCh38
rs525697 Chr.17: 7024129-7024129 on Build GRCh38
rs1054342322 Chr.17: 7024176-7024176 on Build GRCh38
rs546801346 Chr.17: 7024160-7024160 on Build GRCh38
rs369235158 Chr.17: 7024151-7024151 on Build GRCh38
rs768774894 Chr.17: 7024109-7024109 on Build GRCh38
rs779350768 Chr.17: 7024173-7024173 on Build GRCh38
rs759470605 Chr.17: 7024139-7024139 on Build GRCh38
rs758803820 Chr.17: 7024186-7024186 on Build GRCh38
rs368059971 Chr.17: 7024099-7024099 on Build GRCh38
rs982076224 Chr.17: 7024054-7024054 on Build GRCh38
rs550641068 Chr.17: 7024118-7024118 on Build GRCh38
rs986736706 Chr.17: 7024116-7024116 on Build GRCh38
rs761884588 Chr.17: 7024115-7024115 on Build GRCh38
rs115411695 Chr.17: 7024185-7024185 on Build GRCh38

View Full Product Details