Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76618390-76618639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
UniGene
Hs.42502
Gene Chromosome Location
Chr.5: 76615482-76623434 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1438627 c.72A>C p.E24D Chr.5: 76618635-76618635 on Build GRCh38
3617934 c.70G>A p.E24K Chr.5: 76618637-76618637 on Build GRCh38
3617933 c.168G>A p.L56L Chr.5: 76618539-76618539 on Build GRCh38
4966383 c.101C>A p.T34N Chr.5: 76618606-76618606 on Build GRCh38
3769793 c.174C>T p.G58G Chr.5: 76618533-76618533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199782522 Chr.5: 76618434-76618434 on Build GRCh38
rs532074209 Chr.5: 76618441-76618441 on Build GRCh38
rs766905464 Chr.5: 76618565-76618565 on Build GRCh38
rs746928505 Chr.5: 76618534-76618534 on Build GRCh38
rs866440854 Chr.5: 76618554-76618554 on Build GRCh38
rs768017400 Chr.5: 76618477-76618477 on Build GRCh38
rs140813985 Chr.5: 76618573-76618573 on Build GRCh38
rs773386579 Chr.5: 76618528-76618528 on Build GRCh38
rs202236954 Chr.5: 76618586-76618586 on Build GRCh38
rs925552290 Chr.5: 76618557-76618557 on Build GRCh38
rs761888193 Chr.5: 76618431-76618431 on Build GRCh38
rs766915107 Chr.5: 76618580-76618580 on Build GRCh38
rs776246450 Chr.5: 76618590-76618590 on Build GRCh38
rs761605893 Chr.5: 76618552-76618552 on Build GRCh38
rs780269314 Chr.5: 76618512-76618512 on Build GRCh38
rs370863542 Chr.5: 76618516-76618516 on Build GRCh38
rs749677052 Chr.5: 76618518-76618518 on Build GRCh38
rs769824995 Chr.5: 76618531-76618531 on Build GRCh38
rs755031524 Chr.5: 76618547-76618547 on Build GRCh38
rs750007660 Chr.5: 76618572-76618572 on Build GRCh38
rs772121382 Chr.5: 76618525-76618525 on Build GRCh38
rs267600694 Chr.5: 76618455-76618455 on Build GRCh38
rs751043946 Chr.5: 76618459-76618459 on Build GRCh38
rs759571218 Chr.5: 76618444-76618444 on Build GRCh38
rs997108352 Chr.5: 76618493-76618493 on Build GRCh38
rs112811547 Chr.5: 76618418-76618418 on Build GRCh38
rs754163616 Chr.5: 76618585-76618585 on Build GRCh38
rs748180614 Chr.5: 76618521-76618521 on Build GRCh38
rs1028071777 Chr.5: 76618498-76618498 on Build GRCh38
rs776459252 Chr.5: 76618438-76618438 on Build GRCh38
rs762843040 Chr.5: 76618543-76618543 on Build GRCh38
rs764914705 Chr.5: 76618449-76618449 on Build GRCh38
rs372087578 Chr.5: 76618519-76618519 on Build GRCh38
rs200219056 Chr.5: 76618415-76618415 on Build GRCh38
rs745784256 Chr.5: 76618530-76618530 on Build GRCh38
rs373533121 Chr.5: 76618473-76618473 on Build GRCh38
rs886988565 Chr.5: 76618421-76618421 on Build GRCh38
rs763849424 Chr.5: 76618453-76618453 on Build GRCh38
rs371065191 Chr.5: 76618472-76618472 on Build GRCh38
rs376221361 Chr.5: 76618489-76618489 on Build GRCh38
rs750871274 Chr.5: 76618485-76618485 on Build GRCh38
rs147259966 Chr.5: 76618544-76618544 on Build GRCh38
rs746796916 Chr.5: 76618424-76618424 on Build GRCh38
rs548589144 Chr.5: 76618476-76618476 on Build GRCh38
rs368023912 Chr.5: 76618579-76618579 on Build GRCh38
rs1004107141 Chr.5: 76618426-76618426 on Build GRCh38
rs376889876 Chr.5: 76618451-76618451 on Build GRCh38
rs748342168 Chr.5: 76618607-76618607 on Build GRCh38

View Full Product Details