Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151806858-151807093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7258418 c.425delT p.L142fs*1 Chr.2: 151806887-151806887 on Build GRCh38
4085613 c.406G>A p.A136T Chr.2: 151806906-151806906 on Build GRCh38
5677257 c.439A>G p.I147V Chr.2: 151806873-151806873 on Build GRCh38
6604830 c.392A>C p.E131A Chr.2: 151806920-151806920 on Build GRCh38
3694914 c.420G>A p.Q140Q Chr.2: 151806892-151806892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768409682 Chr.2: 151807006-151807006 on Build GRCh38
rs780521220 Chr.2: 151806903-151806903 on Build GRCh38
rs758790180 Chr.2: 151806901-151806901 on Build GRCh38
rs778783352 Chr.2: 151806975-151806975 on Build GRCh38
rs748170373 Chr.2: 151806965-151806965 on Build GRCh38
rs769867613 Chr.2: 151806967-151806967 on Build GRCh38
rs751897647 Chr.2: 151807013-151807014 on Build GRCh38
rs61737675 Chr.2: 151806892-151806892 on Build GRCh38
rs186242807 Chr.2: 151806985-151806985 on Build GRCh38
rs751702231 Chr.2: 151806924-151806924 on Build GRCh38
rs181841961 Chr.2: 151806938-151806938 on Build GRCh38
rs936446596 Chr.2: 151806907-151806907 on Build GRCh38
rs978233735 Chr.2: 151806895-151806895 on Build GRCh38
rs750966947 Chr.2: 151806900-151806900 on Build GRCh38
rs991834532 Chr.2: 151807027-151807027 on Build GRCh38
rs947815609 Chr.2: 151807054-151807054 on Build GRCh38
rs771741783 Chr.2: 151806991-151806991 on Build GRCh38
rs765778672 Chr.2: 151806894-151806894 on Build GRCh38
rs916256369 Chr.2: 151807041-151807041 on Build GRCh38
rs781119879 Chr.2: 151806947-151806947 on Build GRCh38
rs775392664 Chr.2: 151806993-151806993 on Build GRCh38

View Full Product Details