Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130076245-130076518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3896752 c.2693C>A p.P898H Chr.8: 130076356-130076356 on Build GRCh38
7356532 c.2658G>C p.K886N Chr.8: 130076391-130076391 on Build GRCh38
6605422 c.2646C>T p.T882T Chr.8: 130076403-130076403 on Build GRCh38
5873913 c.2684C>T p.P895L Chr.8: 130076365-130076365 on Build GRCh38
4824717 c.2680C>T p.L894F Chr.8: 130076369-130076369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758641875 Chr.8: 130076303-130076303 on Build GRCh38
rs968184644 Chr.8: 130076358-130076358 on Build GRCh38
rs761614574 Chr.8: 130076437-130076437 on Build GRCh38
rs771701018 Chr.8: 130076445-130076445 on Build GRCh38
rs768617655 Chr.8: 130076417-130076417 on Build GRCh38
rs928380743 Chr.8: 130076403-130076403 on Build GRCh38
rs374657898 Chr.8: 130076386-130076386 on Build GRCh38
rs779486286 Chr.8: 130076388-130076388 on Build GRCh38
rs748771388 Chr.8: 130076409-130076409 on Build GRCh38
rs914229649 Chr.8: 130076326-130076326 on Build GRCh38
rs1051247035 Chr.8: 130076292-130076292 on Build GRCh38
rs999229521 Chr.8: 130076406-130076406 on Build GRCh38
rs1022039530 Chr.8: 130076325-130076325 on Build GRCh38
rs1031098123 Chr.8: 130076430-130076430 on Build GRCh38
rs769177787 Chr.8: 130076421-130076421 on Build GRCh38
rs745445316 Chr.8: 130076324-130076324 on Build GRCh38
rs774227116 Chr.8: 130076422-130076422 on Build GRCh38
rs369830284 Chr.8: 130076328-130076328 on Build GRCh38
rs143080840 Chr.8: 130076321-130076321 on Build GRCh38
rs370779716 Chr.8: 130076319-130076319 on Build GRCh38
rs773059361 Chr.8: 130076457-130076457 on Build GRCh38
rs753143166 Chr.8: 130076302-130076302 on Build GRCh38

View Full Product Details