Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45872641-45872914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA3
Entrez Gene ID
3171
Gene Name
forkhead box A3
Gene Aliases
FKHH3, HNF3G, TCF3G
UniGene
Hs.36137
Gene Chromosome Location
Chr.19: 45864260-45873797 on Build GRCh38
Transcripts
NM_004497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
294382 c.697G>A p.G233R Chr.19: 45872702-45872702 on Build GRCh38
4079483 c.838A>G p.T280A Chr.19: 45872843-45872843 on Build GRCh38
7088309 c.639C>T p.F213F Chr.19: 45872644-45872644 on Build GRCh38
4079482 c.665G>A p.G222E Chr.19: 45872670-45872670 on Build GRCh38
5722986 c.698G>A p.G233E Chr.19: 45872703-45872703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772799849 Chr.19: 45872833-45872833 on Build GRCh38
rs760139463 Chr.19: 45872844-45872844 on Build GRCh38
rs778185458 Chr.19: 45872868-45872868 on Build GRCh38
rs531001040 Chr.19: 45872701-45872701 on Build GRCh38
rs28666870 Chr.19: 45872785-45872785 on Build GRCh38
rs769546205 Chr.19: 45872893-45872893 on Build GRCh38
rs762659076 Chr.19: 45872834-45872834 on Build GRCh38
rs776353999 Chr.19: 45872737-45872737 on Build GRCh38
rs770985467 Chr.19: 45872800-45872800 on Build GRCh38
rs1004526212 Chr.19: 45872805-45872805 on Build GRCh38
rs773364143 Chr.19: 45872730-45872730 on Build GRCh38
rs764817302 Chr.19: 45872702-45872702 on Build GRCh38
rs769846056 Chr.19: 45872727-45872727 on Build GRCh38
rs767307934 Chr.19: 45872670-45872670 on Build GRCh38
rs768531867 Chr.19: 45872819-45872819 on Build GRCh38
rs548866684 Chr.19: 45872707-45872707 on Build GRCh38
rs769611712 Chr.19: 45872832-45872832 on Build GRCh38
rs751051964 Chr.19: 45872839-45872839 on Build GRCh38
rs556790003 Chr.19: 45872755-45872755 on Build GRCh38
rs200491833 Chr.19: 45872892-45872892 on Build GRCh38
rs760604416 Chr.19: 45872736-45872736 on Build GRCh38
rs760500600 Chr.19: 45872677-45872677 on Build GRCh38
rs763864928 Chr.19: 45872678-45872678 on Build GRCh38
rs138611202 Chr.19: 45872891-45872891 on Build GRCh38
rs879018916 Chr.19: 45872760-45872760 on Build GRCh38
rs753217438 Chr.19: 45872849-45872849 on Build GRCh38
rs371687599 Chr.19: 45872756-45872756 on Build GRCh38
rs1047679583 Chr.19: 45872767-45872767 on Build GRCh38
rs368851781 Chr.19: 45872878-45872878 on Build GRCh38
rs778898121 Chr.19: 45872801-45872801 on Build GRCh38
rs749657388 Chr.19: 45872721-45872721 on Build GRCh38
rs552726739 Chr.19: 45872798-45872798 on Build GRCh38
rs750062748 Chr.19: 45872754-45872754 on Build GRCh38
rs759526623 Chr.19: 45872665-45872665 on Build GRCh38
rs746963773 Chr.19: 45872812-45872812 on Build GRCh38
rs773095488 Chr.19: 45872664-45872664 on Build GRCh38
rs1019491339 Chr.19: 45872729-45872729 on Build GRCh38
rs1029632389 Chr.19: 45872852-45872852 on Build GRCh38
rs1030143565 Chr.19: 45872696-45872696 on Build GRCh38
rs753059025 Chr.19: 45872769-45872769 on Build GRCh38
rs771010717 Chr.19: 45872733-45872733 on Build GRCh38
rs754013121 Chr.19: 45872869-45872869 on Build GRCh38
rs201747924 Chr.19: 45872748-45872748 on Build GRCh38
rs776446460 Chr.19: 45872826-45872826 on Build GRCh38
rs768096522 Chr.19: 45872845-45872845 on Build GRCh38
rs937004765 Chr.19: 45872743-45872743 on Build GRCh38
rs775453392 Chr.19: 45872735-45872735 on Build GRCh38
rs772989749 Chr.19: 45872894-45872894 on Build GRCh38
rs200743469 Chr.19: 45872716-45872716 on Build GRCh38
rs946058176 Chr.19: 45872890-45872890 on Build GRCh38
rs765990333 Chr.19: 45872836-45872836 on Build GRCh38
rs150110884 Chr.19: 45872850-45872850 on Build GRCh38
rs758978324 Chr.19: 45872708-45872708 on Build GRCh38
rs781697700 Chr.19: 45872766-45872766 on Build GRCh38
rs140760356 Chr.19: 45872831-45872831 on Build GRCh38
rs753556194 Chr.19: 45872680-45872680 on Build GRCh38
rs539294853 Chr.19: 45872797-45872797 on Build GRCh38
rs541734780 Chr.19: 45872879-45872879 on Build GRCh38
rs781684132 Chr.19: 45872722-45872722 on Build GRCh38
rs868733101 Chr.19: 45872793-45872793 on Build GRCh38

View Full Product Details