Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177513320-177513567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX41
Entrez Gene ID
51428
Gene Name
DEAD-box helicase 41
Gene Aliases
ABS, MPLPF
UniGene
Hs.484288
Gene Chromosome Location
Chr.5: 177511577-177517326 on Build GRCh38
Transcripts
NM_001321732.1 NM_001321830.1 NM_016222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3744796 c.1229A>G p.Q410R Chr.5: 177513354-177513354 on Build GRCh38
158650 c.1230+1G>A p.? Chr.5: 177513352-177513352 on Build GRCh38
3761138 c.1200C>T p.R400R Chr.5: 177513383-177513383 on Build GRCh38
234606 c.1131C>T p.T377T Chr.5: 177513452-177513452 on Build GRCh38
3994221 c.1126G>C p.A376P Chr.5: 177513457-177513457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144382614 Chr.5: 177513446-177513446 on Build GRCh38
rs747072227 Chr.5: 177513396-177513396 on Build GRCh38
rs748411098 Chr.5: 177513464-177513464 on Build GRCh38
rs200509082 Chr.5: 177513533-177513533 on Build GRCh38
rs990760376 Chr.5: 177513517-177513517 on Build GRCh38
rs867147632 Chr.5: 177513452-177513452 on Build GRCh38
rs781544959 Chr.5: 177513372-177513372 on Build GRCh38
rs144057590 Chr.5: 177513442-177513442 on Build GRCh38
rs764255682 Chr.5: 177513376-177513376 on Build GRCh38
rs759066983 Chr.5: 177513495-177513495 on Build GRCh38
rs981444008 Chr.5: 177513540-177513540 on Build GRCh38
rs777898451 Chr.5: 177513536-177513536 on Build GRCh38
rs768748580 Chr.5: 177513416-177513416 on Build GRCh38
rs201899140 Chr.5: 177513500-177513500 on Build GRCh38
rs532895566 Chr.5: 177513493-177513493 on Build GRCh38
rs201410094 Chr.5: 177513518-177513518 on Build GRCh38
rs756355310 Chr.5: 177513534-177513534 on Build GRCh38
rs762205641 Chr.5: 177513404-177513405 on Build GRCh38
rs758885853 Chr.5: 177513382-177513382 on Build GRCh38
rs767255662 Chr.5: 177513356-177513356 on Build GRCh38
rs757521888 Chr.5: 177513359-177513359 on Build GRCh38
rs747672157 Chr.5: 177513478-177513478 on Build GRCh38
rs780493149 Chr.5: 177513511-177513511 on Build GRCh38
rs769417886 Chr.5: 177513471-177513471 on Build GRCh38
rs781350513 Chr.5: 177513523-177513523 on Build GRCh38
rs750951330 Chr.5: 177513408-177513409 on Build GRCh38
rs556840837 Chr.5: 177513380-177513380 on Build GRCh38
rs763708504 Chr.5: 177513504-177513504 on Build GRCh38
rs200003905 Chr.5: 177513466-177513466 on Build GRCh38
rs988994238 Chr.5: 177513398-177513398 on Build GRCh38
rs774477210 Chr.5: 177513489-177513489 on Build GRCh38
rs767865139 Chr.5: 177513406-177513407 on Build GRCh38
rs200279634 Chr.5: 177513496-177513496 on Build GRCh38
rs779335524 Chr.5: 177513368-177513368 on Build GRCh38
rs752416266 Chr.5: 177513358-177513358 on Build GRCh38
rs335438 Chr.5: 177513383-177513383 on Build GRCh38
rs756575999 Chr.5: 177513495-177513495 on Build GRCh38
rs997092535 Chr.5: 177513371-177513371 on Build GRCh38
rs751760054 Chr.5: 177513392-177513393 on Build GRCh38
rs773047033 Chr.5: 177513473-177513473 on Build GRCh38
rs545745122 Chr.5: 177513524-177513524 on Build GRCh38
rs750792912 Chr.5: 177513505-177513505 on Build GRCh38
rs774143705 Chr.5: 177513407-177513407 on Build GRCh38
rs758799936 Chr.5: 177513508-177513508 on Build GRCh38
rs959191650 Chr.5: 177513507-177513507 on Build GRCh38
rs752095774 Chr.5: 177513514-177513514 on Build GRCh38

View Full Product Details