Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43363797-43364058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL1
Entrez Gene ID
64834
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
CGI-88, Ssc1
UniGene
Hs.25597
Gene Chromosome Location
Chr.1: 43363397-43368074 on Build GRCh38
Transcripts
NM_001256399.1 NM_001256401.1 NM_001256402.1 NM_022821.3 NR_046117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489891 c.786C>T p.P262P Chr.1: 43363970-43363970 on Build GRCh38
4167693 c.740T>A p.F247Y Chr.1: 43364016-43364016 on Build GRCh38
3369773 c.700C>T p.H234Y Chr.1: 43364056-43364056 on Build GRCh38
1185440 c.779G>T p.R260L Chr.1: 43363977-43363977 on Build GRCh38
5647225 c.736C>A p.L246M Chr.1: 43364020-43364020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781755302 Chr.1: 43363929-43363929 on Build GRCh38
rs780285888 Chr.1: 43363905-43363905 on Build GRCh38
rs147713711 Chr.1: 43364036-43364036 on Build GRCh38
rs970569989 Chr.1: 43363942-43363942 on Build GRCh38
rs200400885 Chr.1: 43363930-43363930 on Build GRCh38
rs749358750 Chr.1: 43363853-43363853 on Build GRCh38
rs747340436 Chr.1: 43363913-43363913 on Build GRCh38
rs773639085 Chr.1: 43363966-43363966 on Build GRCh38
rs372086459 Chr.1: 43364026-43364026 on Build GRCh38
rs753277637 Chr.1: 43364030-43364030 on Build GRCh38
rs763781268 Chr.1: 43363890-43363890 on Build GRCh38
rs761384500 Chr.1: 43364032-43364032 on Build GRCh38
rs183418674 Chr.1: 43363896-43363896 on Build GRCh38
rs775381609 Chr.1: 43363997-43363997 on Build GRCh38
rs748289830 Chr.1: 43363934-43363934 on Build GRCh38
rs896989963 Chr.1: 43363864-43363864 on Build GRCh38
rs768851320 Chr.1: 43364024-43364024 on Build GRCh38
rs755471252 Chr.1: 43363923-43363923 on Build GRCh38
rs752230959 Chr.1: 43363888-43363888 on Build GRCh38
rs1024845378 Chr.1: 43363920-43363920 on Build GRCh38
rs1005460454 Chr.1: 43363834-43363834 on Build GRCh38
rs1009401687 Chr.1: 43363909-43363909 on Build GRCh38
rs772035589 Chr.1: 43363979-43363979 on Build GRCh38
rs759332122 Chr.1: 43363978-43363978 on Build GRCh38
rs544159505 Chr.1: 43363895-43363895 on Build GRCh38
rs532979031 Chr.1: 43363969-43363969 on Build GRCh38
rs11558091 Chr.1: 43364031-43364031 on Build GRCh38
rs774131749 Chr.1: 43363868-43363868 on Build GRCh38
rs759408514 Chr.1: 43363878-43363878 on Build GRCh38
rs780307154 Chr.1: 43363831-43363831 on Build GRCh38
rs755587257 Chr.1: 43363889-43363889 on Build GRCh38
rs760234755 Chr.1: 43364009-43364009 on Build GRCh38
rs1050203567 Chr.1: 43363835-43363835 on Build GRCh38
rs953945590 Chr.1: 43363943-43363943 on Build GRCh38
rs769982072 Chr.1: 43363959-43363959 on Build GRCh38
rs774040282 Chr.1: 43363976-43363976 on Build GRCh38
rs764905013 Chr.1: 43364035-43364035 on Build GRCh38
rs767436294 Chr.1: 43363885-43363885 on Build GRCh38

View Full Product Details