Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73577302-73577575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
UniGene
Hs.496323
Gene Chromosome Location
Chr.X: 73563045-73687109 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029582.1 XM_017029583.1 XM_017029584.1

SNP Detail

SNP ID Chromosome Location
rs760685818 Chr.X: 73577476-73577476 on Build GRCh38
rs2486782 Chr.X: 73577344-73577344 on Build GRCh38
rs775576193 Chr.X: 73577352-73577352 on Build GRCh38
rs376441074 Chr.X: 73577390-73577390 on Build GRCh38
rs373733681 Chr.X: 73577447-73577447 on Build GRCh38
rs772315520 Chr.X: 73577345-73577345 on Build GRCh38
rs141801100 Chr.X: 73577527-73577527 on Build GRCh38
rs764383875 Chr.X: 73577378-73577378 on Build GRCh38
rs780222964 Chr.X: 73577328-73577328 on Build GRCh38
rs765003646 Chr.X: 73577500-73577500 on Build GRCh38
rs934275719 Chr.X: 73577394-73577394 on Build GRCh38
rs201774174 Chr.X: 73577434-73577434 on Build GRCh38
rs752278734 Chr.X: 73577504-73577504 on Build GRCh38
rs754449791 Chr.X: 73577484-73577484 on Build GRCh38
rs200590647 Chr.X: 73577335-73577336 on Build GRCh38
rs864622038 Chr.X: 73577441-73577441 on Build GRCh38
rs763672140 Chr.X: 73577473-73577477 on Build GRCh38
rs112789730 Chr.X: 73577343-73577343 on Build GRCh38
rs373305689 Chr.X: 73577371-73577371 on Build GRCh38
rs1051023468 Chr.X: 73577446-73577446 on Build GRCh38

View Full Product Details