Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38891636-38891909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A1
Entrez Gene ID
92689
Gene Name
family with sequence similarity 114 member A1
Gene Aliases
Noxp20
UniGene
Hs.476517
Gene Chromosome Location
Chr.4: 38867733-38945744 on Build GRCh38
Transcripts
NM_138389.2 NR_033290.1 XM_005262672.2 XM_006714033.2 XM_017008838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545353 c.400G>T p.G134C Chr.4: 38891794-38891794 on Build GRCh38
7283922 c.401G>A p.G134D Chr.4: 38891795-38891795 on Build GRCh38
2846423 c.417G>A p.S139S Chr.4: 38891811-38891811 on Build GRCh38
4625289 c.427G>A p.A143T Chr.4: 38891821-38891821 on Build GRCh38
6658107 c.396C>A p.S132S Chr.4: 38891790-38891790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573835375 Chr.4: 38891755-38891755 on Build GRCh38
rs752288988 Chr.4: 38891783-38891783 on Build GRCh38
rs376471969 Chr.4: 38891701-38891701 on Build GRCh38
rs572671558 Chr.4: 38891810-38891810 on Build GRCh38
rs746057332 Chr.4: 38891793-38891793 on Build GRCh38
rs765616617 Chr.4: 38891863-38891863 on Build GRCh38
rs16994884 Chr.4: 38891674-38891674 on Build GRCh38
rs141631653 Chr.4: 38891756-38891756 on Build GRCh38
rs766125263 Chr.4: 38891740-38891740 on Build GRCh38
rs763458551 Chr.4: 38891881-38891881 on Build GRCh38
rs968442761 Chr.4: 38891821-38891821 on Build GRCh38
rs545106237 Chr.4: 38891788-38891788 on Build GRCh38
rs747046410 Chr.4: 38891773-38891773 on Build GRCh38
rs776720663 Chr.4: 38891776-38891776 on Build GRCh38
rs953948201 Chr.4: 38891720-38891720 on Build GRCh38
rs918358210 Chr.4: 38891792-38891792 on Build GRCh38
rs914192852 Chr.4: 38891815-38891815 on Build GRCh38
rs980240911 Chr.4: 38891828-38891828 on Build GRCh38
rs1032197019 Chr.4: 38891679-38891679 on Build GRCh38
rs774479090 Chr.4: 38891699-38891699 on Build GRCh38
rs780944265 Chr.4: 38891825-38891825 on Build GRCh38
rs753005591 Chr.4: 38891878-38891878 on Build GRCh38
rs766689920 Chr.4: 38891771-38891771 on Build GRCh38
rs199989358 Chr.4: 38891758-38891758 on Build GRCh38
rs770323117 Chr.4: 38891836-38891836 on Build GRCh38
rs781536756 Chr.4: 38891791-38891791 on Build GRCh38
rs777848114 Chr.4: 38891772-38891772 on Build GRCh38
rs956136804 Chr.4: 38891683-38891683 on Build GRCh38
rs760288286 Chr.4: 38891831-38891831 on Build GRCh38
rs898510644 Chr.4: 38891717-38891717 on Build GRCh38
rs776383272 Chr.4: 38891842-38891842 on Build GRCh38
rs753230307 Chr.4: 38891767-38891767 on Build GRCh38
rs147040180 Chr.4: 38891766-38891766 on Build GRCh38
rs541589932 Chr.4: 38891850-38891850 on Build GRCh38
rs773017701 Chr.4: 38891811-38891811 on Build GRCh38
rs988761378 Chr.4: 38891812-38891812 on Build GRCh38
rs201447299 Chr.4: 38891769-38891769 on Build GRCh38
rs371204707 Chr.4: 38891723-38891723 on Build GRCh38
rs375458001 Chr.4: 38891808-38891808 on Build GRCh38
rs757698183 Chr.4: 38891861-38891861 on Build GRCh38
rs760250592 Chr.4: 38891733-38891733 on Build GRCh38
rs1056607129 Chr.4: 38891849-38891849 on Build GRCh38
rs1037418949 Chr.4: 38891702-38891702 on Build GRCh38
rs772721151 Chr.4: 38891726-38891726 on Build GRCh38

View Full Product Details