Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31758493-31758730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC5
Entrez Gene ID
9681
Gene Name
DEP domain containing 5
Gene Aliases
DEP.5, FFEVF
UniGene
Hs.435022
Gene Chromosome Location
Chr.22: 31753951-31907034 on Build GRCh38
Transcripts
NM_001007188.2 NM_001136029.2 NM_001242896.1 NM_001242897.1 NM_014662.4 NR_110988.1 XM_005261862.1 XM_011530557.2 XM_011530558.2 XM_011530559.2 XM_011530560.2 XM_011530561.2 XM_011530562.2 XM_011530563.2 XM_011530565.2 XM_011530568.2 XM_017029109.1 XM_017029110.1 XM_017029111.1 XM_017029112.1 XM_017029113.1 XM_017029114.1 XM_017029115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506245 c.139G>A p.E47K Chr.22: 31758626-31758626 on Build GRCh38
2938598 c.69A>G p.L23L Chr.22: 31758556-31758556 on Build GRCh38
6506246 c.139G>A p.E47K Chr.22: 31758626-31758626 on Build GRCh38
4441030 c.109G>A p.D37N Chr.22: 31758596-31758596 on Build GRCh38
3681419 c.69A>G p.L23L Chr.22: 31758556-31758556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766327865 Chr.22: 31758589-31758589 on Build GRCh38
rs368243595 Chr.22: 31758622-31758622 on Build GRCh38
rs759350041 Chr.22: 31758653-31758653 on Build GRCh38
rs756878843 Chr.22: 31758617-31758617 on Build GRCh38
rs572660873 Chr.22: 31758558-31758558 on Build GRCh38
rs748264035 Chr.22: 31758638-31758638 on Build GRCh38
rs934023877 Chr.22: 31758534-31758534 on Build GRCh38
rs760541660 Chr.22: 31758582-31758582 on Build GRCh38
rs374794334 Chr.22: 31758577-31758577 on Build GRCh38
rs956844670 Chr.22: 31758623-31758623 on Build GRCh38
rs749841723 Chr.22: 31758553-31758553 on Build GRCh38
rs886039243 Chr.22: 31758545-31758545 on Build GRCh38
rs368035159 Chr.22: 31758584-31758584 on Build GRCh38
rs762542455 Chr.22: 31758677-31758677 on Build GRCh38
rs769208150 Chr.22: 31758624-31758624 on Build GRCh38
rs755036534 Chr.22: 31758599-31758599 on Build GRCh38
rs780433877 Chr.22: 31758620-31758620 on Build GRCh38
rs756812375 Chr.22: 31758663-31758663 on Build GRCh38
rs763669717 Chr.22: 31758616-31758616 on Build GRCh38
rs1023389808 Chr.22: 31758600-31758600 on Build GRCh38
rs370544271 Chr.22: 31758593-31758593 on Build GRCh38
rs761019212 Chr.22: 31758644-31758644 on Build GRCh38
rs757783508 Chr.22: 31758615-31758615 on Build GRCh38
rs886039244 Chr.22: 31758619-31758620 on Build GRCh38
rs765274087 Chr.22: 31758654-31758654 on Build GRCh38
rs761783252 Chr.22: 31758563-31758563 on Build GRCh38
rs200119826 Chr.22: 31758556-31758556 on Build GRCh38
rs535630975 Chr.22: 31758625-31758625 on Build GRCh38
rs776627801 Chr.22: 31758647-31758647 on Build GRCh38
rs375173943 Chr.22: 31758614-31758614 on Build GRCh38
rs752540695 Chr.22: 31758670-31758670 on Build GRCh38
rs763558156 Chr.22: 31758684-31758684 on Build GRCh38
rs771997222 Chr.22: 31758642-31758642 on Build GRCh38
rs548953445 Chr.22: 31758645-31758645 on Build GRCh38
rs566463688 Chr.22: 31758601-31758601 on Build GRCh38
rs902191906 Chr.22: 31758682-31758682 on Build GRCh38

View Full Product Details