Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65004146-65004413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419964 c.162G>T p.K54N Chr.14: 65004266-65004266 on Build GRCh38
3497202 c.174C>T p.V58V Chr.14: 65004278-65004278 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368879213 Chr.14: 65004215-65004215 on Build GRCh38
rs868751342 Chr.14: 65004290-65004290 on Build GRCh38
rs772690370 Chr.14: 65004282-65004282 on Build GRCh38
rs766346583 Chr.14: 65004338-65004338 on Build GRCh38
rs765358068 Chr.14: 65004235-65004235 on Build GRCh38
rs768861678 Chr.14: 65004303-65004303 on Build GRCh38
rs762014629 Chr.14: 65004233-65004233 on Build GRCh38
rs763052121 Chr.14: 65004246-65004246 on Build GRCh38
rs775985377 Chr.14: 65004224-65004224 on Build GRCh38
rs1037314116 Chr.14: 65004349-65004349 on Build GRCh38
rs751384349 Chr.14: 65004252-65004252 on Build GRCh38
rs777301110 Chr.14: 65004270-65004270 on Build GRCh38
rs756590934 Chr.14: 65004178-65004178 on Build GRCh38
rs760932142 Chr.14: 65004225-65004225 on Build GRCh38
rs890254294 Chr.14: 65004393-65004393 on Build GRCh38
rs375425851 Chr.14: 65004210-65004210 on Build GRCh38
rs930167829 Chr.14: 65004201-65004201 on Build GRCh38
rs200384517 Chr.14: 65004314-65004314 on Build GRCh38
rs755744657 Chr.14: 65004267-65004267 on Build GRCh38
rs779423198 Chr.14: 65004205-65004205 on Build GRCh38
rs759399892 Chr.14: 65004355-65004355 on Build GRCh38
rs776925757 Chr.14: 65004231-65004231 on Build GRCh38
rs780363660 Chr.14: 65004292-65004292 on Build GRCh38
rs771638373 Chr.14: 65004204-65004204 on Build GRCh38
rs917365184 Chr.14: 65004165-65004165 on Build GRCh38
rs889704103 Chr.14: 65004196-65004196 on Build GRCh38
rs752302231 Chr.14: 65004262-65004262 on Build GRCh38
rs532297504 Chr.14: 65004209-65004209 on Build GRCh38
rs771277535 Chr.14: 65004368-65004368 on Build GRCh38
rs535858782 Chr.14: 65004238-65004238 on Build GRCh38
rs766215092 Chr.14: 65004251-65004251 on Build GRCh38
rs746426903 Chr.14: 65004211-65004211 on Build GRCh38
rs375096435 Chr.14: 65004348-65004348 on Build GRCh38
rs748787797 Chr.14: 65004275-65004275 on Build GRCh38
rs996048056 Chr.14: 65004373-65004373 on Build GRCh38
rs1042997846 Chr.14: 65004183-65004183 on Build GRCh38
rs768934987 Chr.14: 65004226-65004226 on Build GRCh38
rs1051690909 Chr.14: 65004390-65004390 on Build GRCh38
rs777015815 Chr.14: 65004304-65004304 on Build GRCh38
rs560735586 Chr.14: 65004321-65004321 on Build GRCh38
rs770136544 Chr.14: 65004327-65004327 on Build GRCh38
rs774119852 Chr.14: 65004340-65004340 on Build GRCh38
rs769257165 Chr.14: 65004198-65004198 on Build GRCh38
rs941806881 Chr.14: 65004317-65004317 on Build GRCh38
rs762886547 Chr.14: 65004334-65004334 on Build GRCh38
rs574189581 Chr.14: 65004356-65004356 on Build GRCh38
rs762174175 Chr.14: 65004319-65004319 on Build GRCh38
rs185874927 Chr.14: 65004257-65004257 on Build GRCh38

View Full Product Details