Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14751283-14751522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4404231 c.1983T>G p.I661M Chr.19: 14751477-14751477 on Build GRCh38
3529424 c.2012G>A p.G671E Chr.19: 14751448-14751448 on Build GRCh38
6417594 c.2023C>A p.R675S Chr.19: 14751437-14751437 on Build GRCh38
438741 c.1994C>T p.S665F Chr.19: 14751466-14751466 on Build GRCh38
232250 c.2021C>T p.S674F Chr.19: 14751439-14751439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025635334 Chr.19: 14751382-14751382 on Build GRCh38
rs549687894 Chr.19: 14751321-14751321 on Build GRCh38
rs536007948 Chr.19: 14751317-14751317 on Build GRCh38
rs560171435 Chr.19: 14751443-14751443 on Build GRCh38
rs750078097 Chr.19: 14751432-14751432 on Build GRCh38
rs761330448 Chr.19: 14751424-14751424 on Build GRCh38
rs756780184 Chr.19: 14751501-14751501 on Build GRCh38
rs776849746 Chr.19: 14751402-14751402 on Build GRCh38
rs149862660 Chr.19: 14751379-14751379 on Build GRCh38
rs754857376 Chr.19: 14751389-14751389 on Build GRCh38
rs781653637 Chr.19: 14751486-14751486 on Build GRCh38
rs779016111 Chr.19: 14751446-14751446 on Build GRCh38
rs3752187 Chr.19: 14751466-14751466 on Build GRCh38
rs150922672 Chr.19: 14751456-14751456 on Build GRCh38
rs773600551 Chr.19: 14751397-14751397 on Build GRCh38
rs760159587 Chr.19: 14751411-14751411 on Build GRCh38
rs201548088 Chr.19: 14751412-14751412 on Build GRCh38
rs143530914 Chr.19: 14751494-14751494 on Build GRCh38
rs771057297 Chr.19: 14751469-14751469 on Build GRCh38
rs369479601 Chr.19: 14751353-14751353 on Build GRCh38
rs756311370 Chr.19: 14751461-14751461 on Build GRCh38
rs200792971 Chr.19: 14751413-14751413 on Build GRCh38
rs753569390 Chr.19: 14751438-14751438 on Build GRCh38
rs3752186 Chr.19: 14751393-14751393 on Build GRCh38
rs199986229 Chr.19: 14751401-14751401 on Build GRCh38
rs370157037 Chr.19: 14751503-14751503 on Build GRCh38
rs144670525 Chr.19: 14751436-14751436 on Build GRCh38
rs142522626 Chr.19: 14751435-14751435 on Build GRCh38
rs934516579 Chr.19: 14751352-14751352 on Build GRCh38
rs779201751 Chr.19: 14751391-14751391 on Build GRCh38
rs367674615 Chr.19: 14751450-14751450 on Build GRCh38
rs893094186 Chr.19: 14751354-14751354 on Build GRCh38
rs901537281 Chr.19: 14751410-14751410 on Build GRCh38
rs762685687 Chr.19: 14751376-14751376 on Build GRCh38
rs200022474 Chr.19: 14751454-14751454 on Build GRCh38
rs766971116 Chr.19: 14751431-14751431 on Build GRCh38
rs949408320 Chr.19: 14751312-14751312 on Build GRCh38

View Full Product Details