Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 39626202-39626473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD2
Entrez Gene ID
225
Gene Name
ATP binding cassette subfamily D member 2
Gene Aliases
ABC39, ALDL1, ALDR, ALDRP, hALDR
UniGene
Hs.117852
Gene Chromosome Location
Chr.12: 39530966-39651333 on Build GRCh38
Transcripts
XM_011538027.2 NM_001031748.3 NM_001319247.1 NR_135051.1 XM_005268806.3

Gene Symbol
C12orf40
Entrez Gene ID
283461
Gene Name
chromosome 12 open reading frame 40
Gene Aliases
HEL-206, HEL-S-94
UniGene
Hs.648205
Gene Chromosome Location
Chr.12: 39626167-39908300 on Build GRCh38
Transcripts
XM_011538027.2 NM_001031748.3 NM_001319247.1 NR_135051.1 XM_005268806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628574 c.23+3_23+4delAA p.? Chr.12: 39626362-39626363 on Build GRCh38
6520128 c.22C>A p.R8R Chr.12: 39626358-39626358 on Build GRCh38
4666711 c.23G>A p.R8Q Chr.12: 39626359-39626359 on Build GRCh38
1283713 c.13G>A p.G5R Chr.12: 39626349-39626349 on Build GRCh38
3460427 c.12C>T p.V4V Chr.12: 39626348-39626348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146254875 Chr.12: 39626396-39626396 on Build GRCh38
rs767752703 Chr.12: 39626362-39626362 on Build GRCh38
rs780135927 Chr.12: 39626287-39626287 on Build GRCh38
rs954181441 Chr.12: 39626222-39626222 on Build GRCh38
rs780541124 Chr.12: 39626348-39626349 on Build GRCh38
rs545680587 Chr.12: 39626289-39626289 on Build GRCh38
rs531757494 Chr.12: 39626405-39626405 on Build GRCh38
rs746518733 Chr.12: 39626310-39626310 on Build GRCh38
rs370787745 Chr.12: 39626317-39626317 on Build GRCh38
rs370917720 Chr.12: 39626409-39626409 on Build GRCh38
rs760131165 Chr.12: 39626320-39626320 on Build GRCh38
rs770344364 Chr.12: 39626402-39626402 on Build GRCh38
rs898244546 Chr.12: 39626269-39626269 on Build GRCh38
rs754967724 Chr.12: 39626369-39626369 on Build GRCh38
rs746717078 Chr.12: 39626390-39626390 on Build GRCh38
rs751896348 Chr.12: 39626354-39626354 on Build GRCh38
rs935260093 Chr.12: 39626226-39626226 on Build GRCh38
rs547898997 Chr.12: 39626448-39626448 on Build GRCh38
rs1044869438 Chr.12: 39626252-39626252 on Build GRCh38
rs992512013 Chr.12: 39626433-39626433 on Build GRCh38
rs977207871 Chr.12: 39626258-39626258 on Build GRCh38
rs529342747 Chr.12: 39626423-39626423 on Build GRCh38
rs373587733 Chr.12: 39626303-39626303 on Build GRCh38
rs74088775 Chr.12: 39626313-39626313 on Build GRCh38
rs549722599 Chr.12: 39626373-39626373 on Build GRCh38
rs947730892 Chr.12: 39626238-39626238 on Build GRCh38
rs917595277 Chr.12: 39626231-39626231 on Build GRCh38
rs182652958 Chr.12: 39626282-39626282 on Build GRCh38
rs759857305 Chr.12: 39626445-39626445 on Build GRCh38
rs763205830 Chr.12: 39626340-39626340 on Build GRCh38
rs368689020 Chr.12: 39626364-39626364 on Build GRCh38
rs965820709 Chr.12: 39626254-39626254 on Build GRCh38
rs757398186 Chr.12: 39626359-39626359 on Build GRCh38
rs758256564 Chr.12: 39626385-39626385 on Build GRCh38
rs775942378 Chr.12: 39626334-39626334 on Build GRCh38
rs373435231 Chr.12: 39626392-39626392 on Build GRCh38
rs764411627 Chr.12: 39626346-39626346 on Build GRCh38
rs746309053 Chr.12: 39626400-39626400 on Build GRCh38
rs1021542882 Chr.12: 39626370-39626370 on Build GRCh38
rs369126266 Chr.12: 39626290-39626290 on Build GRCh38
rs887253965 Chr.12: 39626295-39626295 on Build GRCh38
rs751649239 Chr.12: 39626422-39626422 on Build GRCh38
rs747708500 Chr.12: 39626294-39626294 on Build GRCh38
rs749304907 Chr.12: 39626288-39626288 on Build GRCh38
rs992068139 Chr.12: 39626228-39626228 on Build GRCh38
rs752542899 Chr.12: 39626384-39626384 on Build GRCh38
rs772643700 Chr.12: 39626307-39626307 on Build GRCh38
rs747710095 Chr.12: 39626377-39626377 on Build GRCh38
rs765476441 Chr.12: 39626331-39626331 on Build GRCh38

View Full Product Details