Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97910504-97910735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624344 c.470A>T p.K157M Chr.9: 97910556-97910556 on Build GRCh38
6476459 c.507G>T p.V169V Chr.9: 97910519-97910519 on Build GRCh38
199425 c.498G>A p.P166P Chr.9: 97910528-97910528 on Build GRCh38
1314304 c.484G>A p.E162K Chr.9: 97910542-97910542 on Build GRCh38
1103060 c.427T>C p.S143P Chr.9: 97910599-97910599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752584462 Chr.9: 97910558-97910558 on Build GRCh38
rs752531496 Chr.9: 97910615-97910615 on Build GRCh38
rs201767472 Chr.9: 97910594-97910594 on Build GRCh38
rs770487687 Chr.9: 97910585-97910585 on Build GRCh38
rs993737431 Chr.9: 97910592-97910592 on Build GRCh38
rs755499799 Chr.9: 97910625-97910625 on Build GRCh38
rs772887956 Chr.9: 97910540-97910540 on Build GRCh38
rs756096445 Chr.9: 97910560-97910560 on Build GRCh38
rs770291743 Chr.9: 97910660-97910660 on Build GRCh38
rs984883401 Chr.9: 97910547-97910547 on Build GRCh38
rs934983219 Chr.9: 97910706-97910706 on Build GRCh38
rs754731854 Chr.9: 97910553-97910553 on Build GRCh38
rs545348756 Chr.9: 97910695-97910695 on Build GRCh38
rs537572715 Chr.9: 97910614-97910614 on Build GRCh38
rs183394712 Chr.9: 97910606-97910606 on Build GRCh38
rs747230242 Chr.9: 97910651-97910651 on Build GRCh38
rs773786271 Chr.9: 97910662-97910662 on Build GRCh38
rs777236790 Chr.9: 97910527-97910527 on Build GRCh38
rs148247656 Chr.9: 97910591-97910591 on Build GRCh38
rs746015775 Chr.9: 97910563-97910563 on Build GRCh38
rs373301254 Chr.9: 97910529-97910529 on Build GRCh38
rs190841159 Chr.9: 97910554-97910554 on Build GRCh38
rs533167180 Chr.9: 97910676-97910676 on Build GRCh38
rs751180435 Chr.9: 97910550-97910550 on Build GRCh38
rs755195563 Chr.9: 97910657-97910657 on Build GRCh38
rs372941889 Chr.9: 97910631-97910631 on Build GRCh38
rs781686721 Chr.9: 97910658-97910658 on Build GRCh38
rs200779838 Chr.9: 97910580-97910580 on Build GRCh38
rs562591248 Chr.9: 97910675-97910675 on Build GRCh38
rs758783430 Chr.9: 97910548-97910548 on Build GRCh38
rs560191640 Chr.9: 97910698-97910698 on Build GRCh38
rs774019223 Chr.9: 97910610-97910610 on Build GRCh38
rs780265873 Chr.9: 97910567-97910567 on Build GRCh38
rs902268496 Chr.9: 97910686-97910686 on Build GRCh38
rs756043264 Chr.9: 97910620-97910620 on Build GRCh38
rs759137962 Chr.9: 97910611-97910611 on Build GRCh38
rs762686677 Chr.9: 97910544-97910544 on Build GRCh38
rs767250835 Chr.9: 97910557-97910557 on Build GRCh38
rs571150996 Chr.9: 97910528-97910528 on Build GRCh38
rs778934572 Chr.9: 97910562-97910562 on Build GRCh38
rs748578010 Chr.9: 97910659-97910659 on Build GRCh38
rs150857267 Chr.9: 97910570-97910570 on Build GRCh38
rs758536083 Chr.9: 97910565-97910565 on Build GRCh38
rs780019876 Chr.9: 97910641-97910641 on Build GRCh38
rs760594016 Chr.9: 97910640-97910640 on Build GRCh38
rs765808100 Chr.9: 97910602-97910602 on Build GRCh38
rs377088253 Chr.9: 97910707-97910707 on Build GRCh38
rs753786534 Chr.9: 97910632-97910632 on Build GRCh38
rs558175701 Chr.9: 97910569-97910569 on Build GRCh38
rs749786997 Chr.9: 97910665-97910665 on Build GRCh38

View Full Product Details