Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102829315-102829546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
UniGene
Hs.459106
Gene Chromosome Location
Chr.8: 102826302-102864200 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Gene Symbol
LOC107986873
Entrez Gene ID
107986873
Gene Name
forkhead box protein L2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 102810302-102863067 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441789 c.1084G>C p.D362H Chr.8: 102829423-102829423 on Build GRCh38
3762961 c.1026C>T p.Y342Y Chr.8: 102829481-102829481 on Build GRCh38
5503391 c.1134G>A p.W378* Chr.8: 102829373-102829373 on Build GRCh38
6609186 c.1128A>G p.G376G Chr.8: 102829379-102829379 on Build GRCh38
1094432 c.1163C>A p.S388Y Chr.8: 102829344-102829344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748903540 Chr.8: 102829430-102829430 on Build GRCh38
rs781130838 Chr.8: 102829504-102829504 on Build GRCh38
rs375497839 Chr.8: 102829393-102829393 on Build GRCh38
rs769529259 Chr.8: 102829426-102829426 on Build GRCh38
rs141624122 Chr.8: 102829451-102829451 on Build GRCh38
rs145303628 Chr.8: 102829460-102829460 on Build GRCh38
rs754941062 Chr.8: 102829386-102829386 on Build GRCh38
rs772688565 Chr.8: 102829445-102829445 on Build GRCh38
rs764727057 Chr.8: 102829358-102829358 on Build GRCh38
rs780126885 Chr.8: 102829364-102829364 on Build GRCh38
rs906940243 Chr.8: 102829424-102829424 on Build GRCh38
rs113116933 Chr.8: 102829403-102829403 on Build GRCh38
rs772881104 Chr.8: 102829429-102829429 on Build GRCh38
rs766437768 Chr.8: 102829489-102829489 on Build GRCh38
rs372529304 Chr.8: 102829387-102829387 on Build GRCh38
rs62522600 Chr.8: 102829481-102829481 on Build GRCh38
rs747916540 Chr.8: 102829388-102829388 on Build GRCh38
rs758635413 Chr.8: 102829485-102829485 on Build GRCh38
rs755906469 Chr.8: 102829511-102829511 on Build GRCh38
rs746979334 Chr.8: 102829367-102829367 on Build GRCh38
rs748037562 Chr.8: 102829509-102829509 on Build GRCh38
rs777179679 Chr.8: 102829476-102829476 on Build GRCh38
rs369356623 Chr.8: 102829434-102829434 on Build GRCh38
rs765633636 Chr.8: 102829480-102829480 on Build GRCh38

View Full Product Details