Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9680804-9681031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2D
Entrez Gene ID
29121
Gene Name
C-type lectin domain family 2 member D
Gene Aliases
CLAX, LLT1, OCIL
UniGene
Hs.268326
Gene Chromosome Location
Chr.12: 9669708-9699555 on Build GRCh38
Transcripts
NM_001004419.4 NM_001197317.2 NM_001197318.2 NM_001197319.2 NM_013269.5 NR_036693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426873 c.62-3C>G p.? Chr.12: 9680920-9680920 on Build GRCh38
5734627 c.142A>G p.I48V Chr.12: 9681003-9681003 on Build GRCh38
7361495 c.67C>G p.L23V Chr.12: 9680928-9680928 on Build GRCh38
1705135 c.155T>G p.M52R Chr.12: 9681016-9681016 on Build GRCh38
4426874 c.62-3C>G p.? Chr.12: 9680920-9680920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751113726 Chr.12: 9680965-9680965 on Build GRCh38
rs1021756014 Chr.12: 9680899-9680899 on Build GRCh38
rs764588964 Chr.12: 9680894-9680894 on Build GRCh38
rs781379116 Chr.12: 9680956-9680956 on Build GRCh38
rs751427457 Chr.12: 9681008-9681008 on Build GRCh38
rs781138390 Chr.12: 9680913-9680913 on Build GRCh38
rs746072868 Chr.12: 9680958-9680958 on Build GRCh38
rs766013623 Chr.12: 9680922-9680922 on Build GRCh38
rs925054806 Chr.12: 9680978-9680978 on Build GRCh38
rs10844424 Chr.12: 9680830-9680830 on Build GRCh38
rs746747878 Chr.12: 9680976-9680981 on Build GRCh38
rs775886779 Chr.12: 9680889-9680889 on Build GRCh38
rs201704501 Chr.12: 9680968-9680968 on Build GRCh38
rs759258278 Chr.12: 9680981-9680981 on Build GRCh38
rs892148226 Chr.12: 9681002-9681002 on Build GRCh38
rs3764022 Chr.12: 9680928-9680928 on Build GRCh38
rs745528390 Chr.12: 9680932-9680932 on Build GRCh38
rs755895493 Chr.12: 9680953-9680953 on Build GRCh38
rs553694588 Chr.12: 9680934-9680934 on Build GRCh38
rs752486917 Chr.12: 9680988-9680988 on Build GRCh38
rs749890803 Chr.12: 9680912-9680912 on Build GRCh38
rs753403199 Chr.12: 9680926-9680926 on Build GRCh38
rs145574510 Chr.12: 9680964-9680964 on Build GRCh38
rs758349003 Chr.12: 9680931-9680931 on Build GRCh38
rs548316925 Chr.12: 9680886-9680886 on Build GRCh38
rs760123895 Chr.12: 9680919-9680919 on Build GRCh38
rs376493550 Chr.12: 9680875-9680875 on Build GRCh38
rs764990022 Chr.12: 9680984-9680984 on Build GRCh38
rs776263195 Chr.12: 9680975-9680975 on Build GRCh38
rs140143530 Chr.12: 9681003-9681003 on Build GRCh38
rs748204713 Chr.12: 9680932-9680933 on Build GRCh38
rs769100213 Chr.12: 9680970-9680970 on Build GRCh38
rs764055539 Chr.12: 9681007-9681007 on Build GRCh38
rs146151995 Chr.12: 9680971-9680971 on Build GRCh38
rs922002547 Chr.12: 9680999-9680999 on Build GRCh38
rs373724798 Chr.12: 9680891-9680891 on Build GRCh38

View Full Product Details