Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16519052-16519313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_032207.3 XM_006722920.3 NM_006387.5

Gene Symbol
CHERP
Entrez Gene ID
10523
Gene Name
calcium homeostasis endoplasmic reticulum protein
Gene Aliases
DAN16, SCAF6, SRA1
UniGene
Hs.740364
Gene Chromosome Location
Chr.19: 16517889-16542452 on Build GRCh38
Transcripts
NM_032207.3 XM_006722920.3 NM_006387.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628314 c.2630G>A p.R877Q Chr.19: 16519280-16519280 on Build GRCh38
3530031 c.2667T>A p.A889A Chr.19: 16519243-16519243 on Build GRCh38
4773274 c.2722C>A p.R908S Chr.19: 16519188-16519188 on Build GRCh38
6557589 c.2693G>A p.R898H Chr.19: 16519217-16519217 on Build GRCh38
4075167 c.2661C>G p.G887G Chr.19: 16519249-16519249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746597113 Chr.19: 16519192-16519192 on Build GRCh38
rs987852836 Chr.19: 16519092-16519092 on Build GRCh38
rs867218215 Chr.19: 16519176-16519176 on Build GRCh38
rs753404597 Chr.19: 16519151-16519151 on Build GRCh38
rs759185951 Chr.19: 16519219-16519219 on Build GRCh38
rs563213714 Chr.19: 16519145-16519145 on Build GRCh38
rs771072526 Chr.19: 16519134-16519145 on Build GRCh38
rs543574733 Chr.19: 16519138-16519138 on Build GRCh38
rs767656299 Chr.19: 16519157-16519157 on Build GRCh38
rs573469903 Chr.19: 16519113-16519113 on Build GRCh38
rs755230630 Chr.19: 16519177-16519177 on Build GRCh38
rs750997201 Chr.19: 16519167-16519167 on Build GRCh38
rs200671301 Chr.19: 16519126-16519126 on Build GRCh38
rs1036523480 Chr.19: 16519146-16519146 on Build GRCh38
rs118156403 Chr.19: 16519101-16519101 on Build GRCh38
rs946487192 Chr.19: 16519109-16519109 on Build GRCh38
rs749596423 Chr.19: 16519133-16519133 on Build GRCh38
rs757276880 Chr.19: 16519122-16519122 on Build GRCh38
rs372244796 Chr.19: 16519201-16519201 on Build GRCh38
rs764198057 Chr.19: 16519117-16519117 on Build GRCh38
rs766537317 Chr.19: 16519142-16519142 on Build GRCh38
rs368649683 Chr.19: 16519258-16519258 on Build GRCh38
rs967776134 Chr.19: 16519273-16519273 on Build GRCh38
rs370857958 Chr.19: 16519188-16519188 on Build GRCh38
rs775282607 Chr.19: 16519249-16519249 on Build GRCh38
rs777779403 Chr.19: 16519124-16519139 on Build GRCh38
rs545497880 Chr.19: 16519148-16519148 on Build GRCh38
rs374260736 Chr.19: 16519255-16519255 on Build GRCh38
rs756725468 Chr.19: 16519168-16519168 on Build GRCh38
rs370455075 Chr.19: 16519267-16519267 on Build GRCh38
rs781764695 Chr.19: 16519124-16519124 on Build GRCh38
rs201274229 Chr.19: 16519284-16519284 on Build GRCh38
rs780722415 Chr.19: 16519229-16519229 on Build GRCh38
rs747086336 Chr.19: 16519134-16519134 on Build GRCh38
rs749109211 Chr.19: 16519254-16519254 on Build GRCh38
rs745769035 Chr.19: 16519140-16519140 on Build GRCh38
rs528371549 Chr.19: 16519171-16519171 on Build GRCh38
rs773056888 Chr.19: 16519075-16519075 on Build GRCh38
rs374476634 Chr.19: 16519139-16519139 on Build GRCh38
rs375299260 Chr.19: 16519291-16519291 on Build GRCh38
rs973569404 Chr.19: 16519119-16519119 on Build GRCh38
rs199924064 Chr.19: 16519135-16519135 on Build GRCh38
rs374368678 Chr.19: 16519141-16519141 on Build GRCh38
rs751715851 Chr.19: 16519120-16519120 on Build GRCh38
rs970568134 Chr.19: 16519118-16519118 on Build GRCh38
rs553300537 Chr.19: 16519112-16519112 on Build GRCh38
rs767815823 Chr.19: 16519278-16519278 on Build GRCh38
rs532201910 Chr.19: 16519147-16519147 on Build GRCh38
rs542497340 Chr.19: 16519125-16519125 on Build GRCh38
rs769777301 Chr.19: 16519248-16519248 on Build GRCh38
rs771816080 Chr.19: 16519134-16519134 on Build GRCh38
rs374868976 Chr.19: 16519281-16519281 on Build GRCh38
rs780524358 Chr.19: 16519131-16519131 on Build GRCh38
rs754297067 Chr.19: 16519174-16519174 on Build GRCh38
rs536006274 Chr.19: 16519228-16519228 on Build GRCh38
rs956528478 Chr.19: 16519077-16519077 on Build GRCh38
rs370402073 Chr.19: 16519136-16519136 on Build GRCh38
rs61729416 Chr.19: 16519285-16519285 on Build GRCh38
rs201942283 Chr.19: 16519231-16519231 on Build GRCh38
rs377742055 Chr.19: 16519156-16519156 on Build GRCh38

View Full Product Details