Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81672386-81672633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC137
Entrez Gene ID
339230
Gene Name
coiled-coil domain containing 137
Gene Aliases
RaRF
UniGene
Hs.744125
Gene Chromosome Location
Chr.17: 81666697-81673907 on Build GRCh38
Transcripts
NM_199287.2 XM_011524738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821003 c.784T>C p.Y262H Chr.17: 81672618-81672618 on Build GRCh38
5493666 c.792G>A p.A264A Chr.17: 81672626-81672626 on Build GRCh38
708646 c.694C>G p.L232V Chr.17: 81672528-81672528 on Build GRCh38
6417575 c.728C>A p.A243D Chr.17: 81672562-81672562 on Build GRCh38
1387332 c.685C>T p.R229W Chr.17: 81672519-81672519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966906414 Chr.17: 81672429-81672429 on Build GRCh38
rs778907421 Chr.17: 81672613-81672613 on Build GRCh38
rs988675305 Chr.17: 81672535-81672535 on Build GRCh38
rs371268602 Chr.17: 81672559-81672559 on Build GRCh38
rs370963207 Chr.17: 81672507-81672507 on Build GRCh38
rs139696300 Chr.17: 81672445-81672445 on Build GRCh38
rs779868293 Chr.17: 81672534-81672534 on Build GRCh38
rs1022695646 Chr.17: 81672600-81672600 on Build GRCh38
rs1057478132 Chr.17: 81672411-81672411 on Build GRCh38
rs202091713 Chr.17: 81672509-81672509 on Build GRCh38
rs753648466 Chr.17: 81672604-81672604 on Build GRCh38
rs777978705 Chr.17: 81672573-81672573 on Build GRCh38
rs890286699 Chr.17: 81672545-81672545 on Build GRCh38
rs75933860 Chr.17: 81672560-81672560 on Build GRCh38
rs895967843 Chr.17: 81672418-81672418 on Build GRCh38
rs913150471 Chr.17: 81672544-81672544 on Build GRCh38
rs149789072 Chr.17: 81672465-81672465 on Build GRCh38
rs549888801 Chr.17: 81672561-81672561 on Build GRCh38
rs779028277 Chr.17: 81672557-81672557 on Build GRCh38
rs867466889 Chr.17: 81672508-81672508 on Build GRCh38
rs868744173 Chr.17: 81672495-81672495 on Build GRCh38
rs770218523 Chr.17: 81672574-81672574 on Build GRCh38
rs755790449 Chr.17: 81672532-81672532 on Build GRCh38
rs750078935 Chr.17: 81672526-81672526 on Build GRCh38
rs897254986 Chr.17: 81672546-81672546 on Build GRCh38
rs1036493982 Chr.17: 81672609-81672609 on Build GRCh38
rs376394338 Chr.17: 81672594-81672596 on Build GRCh38
rs776931304 Chr.17: 81672567-81672567 on Build GRCh38
rs752673813 Chr.17: 81672615-81672615 on Build GRCh38
rs754827508 Chr.17: 81672611-81672611 on Build GRCh38
rs917355697 Chr.17: 81672588-81672588 on Build GRCh38
rs775991056 Chr.17: 81672579-81672579 on Build GRCh38
rs749942944 Chr.17: 81672583-81672583 on Build GRCh38
rs35963091 Chr.17: 81672594-81672594 on Build GRCh38
rs765755363 Chr.17: 81672515-81672515 on Build GRCh38
rs1005490746 Chr.17: 81672423-81672423 on Build GRCh38
rs11546630 Chr.17: 81672520-81672520 on Build GRCh38
rs144522867 Chr.17: 81672482-81672483 on Build GRCh38
rs1047706770 Chr.17: 81672542-81672542 on Build GRCh38
rs34191669 Chr.17: 81672587-81672589 on Build GRCh38
rs902548396 Chr.17: 81672608-81672608 on Build GRCh38
rs372242681 Chr.17: 81672581-81672581 on Build GRCh38
rs368515571 Chr.17: 81672470-81672470 on Build GRCh38
rs749149362 Chr.17: 81672536-81672536 on Build GRCh38
rs542496439 Chr.17: 81672449-81672449 on Build GRCh38
rs371084134 Chr.17: 81672519-81672519 on Build GRCh38
rs751028100 Chr.17: 81672516-81672516 on Build GRCh38
rs940866869 Chr.17: 81672593-81672593 on Build GRCh38
rs535519979 Chr.17: 81672580-81672580 on Build GRCh38
rs760357623 Chr.17: 81672585-81672585 on Build GRCh38
rs766757722 Chr.17: 81672478-81672478 on Build GRCh38
rs771106380 Chr.17: 81672565-81672565 on Build GRCh38
rs754917458 Chr.17: 81672552-81672552 on Build GRCh38
rs777016790 Chr.17: 81672490-81672490 on Build GRCh38
rs201836165 Chr.17: 81672563-81672563 on Build GRCh38
rs34921873 Chr.17: 81672601-81672602 on Build GRCh38
rs766156952 Chr.17: 81672595-81672595 on Build GRCh38

View Full Product Details