Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37719858-37720103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA10
Entrez Gene ID
284656
Gene Name
EPH receptor A10
Gene Aliases
-
UniGene
Hs.129435
Gene Chromosome Location
Chr.1: 37713888-37765152 on Build GRCh38
Transcripts
NM_001099439.1 XM_017001080.1 XM_017001081.1

SNP Detail

SNP ID Chromosome Location
rs373625157 Chr.1: 37719986-37719986 on Build GRCh38
rs760341550 Chr.1: 37719897-37719897 on Build GRCh38
rs201523562 Chr.1: 37719899-37719899 on Build GRCh38
rs748872840 Chr.1: 37719909-37719909 on Build GRCh38
rs376831140 Chr.1: 37719995-37719995 on Build GRCh38
rs780310300 Chr.1: 37720032-37720032 on Build GRCh38
rs763558088 Chr.1: 37719898-37719898 on Build GRCh38
rs778930454 Chr.1: 37720028-37720028 on Build GRCh38
rs376540009 Chr.1: 37720031-37720031 on Build GRCh38
rs192256218 Chr.1: 37720025-37720025 on Build GRCh38
rs767209870 Chr.1: 37720068-37720068 on Build GRCh38
rs751320707 Chr.1: 37720004-37720004 on Build GRCh38
rs773435840 Chr.1: 37720078-37720078 on Build GRCh38
rs143886883 Chr.1: 37720052-37720052 on Build GRCh38
rs199748160 Chr.1: 37719987-37719987 on Build GRCh38
rs752904524 Chr.1: 37719931-37719931 on Build GRCh38
rs2170809 Chr.1: 37719975-37719975 on Build GRCh38
rs369709653 Chr.1: 37719993-37719993 on Build GRCh38
rs6670599 Chr.1: 37720051-37720051 on Build GRCh38
rs77852596 Chr.1: 37719885-37719885 on Build GRCh38
rs368721119 Chr.1: 37719918-37719918 on Build GRCh38
rs759550839 Chr.1: 37720082-37720082 on Build GRCh38
rs762954604 Chr.1: 37719998-37719998 on Build GRCh38
rs760839982 Chr.1: 37720006-37720006 on Build GRCh38
rs759432555 Chr.1: 37720054-37720054 on Build GRCh38
rs769469649 Chr.1: 37720044-37720044 on Build GRCh38
rs750260220 Chr.1: 37719913-37719913 on Build GRCh38
rs762026616 Chr.1: 37719997-37719997 on Build GRCh38
rs200984509 Chr.1: 37719957-37719957 on Build GRCh38
rs779452079 Chr.1: 37719960-37719960 on Build GRCh38
rs776950931 Chr.1: 37720071-37720071 on Build GRCh38
rs772828546 Chr.1: 37720046-37720046 on Build GRCh38
rs765204480 Chr.1: 37719908-37719908 on Build GRCh38
rs746367810 Chr.1: 37719964-37719964 on Build GRCh38
rs200610563 Chr.1: 37719904-37719904 on Build GRCh38
rs768211661 Chr.1: 37720038-37720038 on Build GRCh38
rs780958682 Chr.1: 37720041-37720041 on Build GRCh38
rs775187156 Chr.1: 37719891-37719891 on Build GRCh38
rs761332888 Chr.1: 37719992-37719992 on Build GRCh38
rs750974576 Chr.1: 37720029-37720029 on Build GRCh38
rs757393344 Chr.1: 37720026-37720026 on Build GRCh38
rs780441627 Chr.1: 37719974-37719974 on Build GRCh38
rs371694877 Chr.1: 37719938-37719938 on Build GRCh38
rs765534268 Chr.1: 37720017-37720018 on Build GRCh38
rs201339817 Chr.1: 37720042-37720042 on Build GRCh38
rs187414532 Chr.1: 37719965-37719965 on Build GRCh38
rs766274598 Chr.1: 37719923-37719923 on Build GRCh38
rs772462490 Chr.1: 37719971-37719971 on Build GRCh38
rs766327882 Chr.1: 37720000-37720000 on Build GRCh38
rs749259623 Chr.1: 37720050-37720050 on Build GRCh38

View Full Product Details