Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167026444-167026705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1OP
Entrez Gene ID
11116
Gene Name
FGFR1 oncogene partner
Gene Aliases
FOP
UniGene
Hs.487175
Gene Chromosome Location
Chr.6: 166999317-167042418 on Build GRCh38
Transcripts
NM_001278690.1 NM_007045.3 NM_194429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4160361 c.929G>C p.G310A Chr.6: 167026556-167026556 on Build GRCh38
5769016 c.983G>C p.G328A Chr.6: 167026610-167026610 on Build GRCh38
1659448 c.961A>G p.S321G Chr.6: 167026588-167026588 on Build GRCh38
377701 c.925A>G p.R309G Chr.6: 167026552-167026552 on Build GRCh38
6207071 c.951G>A p.L317L Chr.6: 167026578-167026578 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764048848 Chr.6: 167026606-167026606 on Build GRCh38
rs768838063 Chr.6: 167026552-167026552 on Build GRCh38
rs144532341 Chr.6: 167026472-167026472 on Build GRCh38
rs1035985260 Chr.6: 167026503-167026503 on Build GRCh38
rs963433343 Chr.6: 167026555-167026555 on Build GRCh38
rs756729225 Chr.6: 167026614-167026614 on Build GRCh38
rs765053714 Chr.6: 167026594-167026594 on Build GRCh38
rs751013217 Chr.6: 167026607-167026607 on Build GRCh38
rs763638175 Chr.6: 167026505-167026505 on Build GRCh38
rs377066763 Chr.6: 167026630-167026630 on Build GRCh38
rs750110428 Chr.6: 167026631-167026631 on Build GRCh38
rs2282861 Chr.6: 167026640-167026640 on Build GRCh38
rs768998367 Chr.6: 167026502-167026502 on Build GRCh38
rs756516294 Chr.6: 167026634-167026634 on Build GRCh38
rs777305495 Chr.6: 167026562-167026562 on Build GRCh38
rs368901489 Chr.6: 167026646-167026646 on Build GRCh38
rs944930285 Chr.6: 167026626-167026626 on Build GRCh38
rs780710798 Chr.6: 167026529-167026529 on Build GRCh38
rs756608056 Chr.6: 167026539-167026539 on Build GRCh38
rs751049665 Chr.6: 167026508-167026508 on Build GRCh38
rs373628095 Chr.6: 167026642-167026642 on Build GRCh38
rs759367211 Chr.6: 167026589-167026589 on Build GRCh38
rs780618516 Chr.6: 167026639-167026639 on Build GRCh38
rs775379729 Chr.6: 167026602-167026602 on Build GRCh38
rs372147195 Chr.6: 167026581-167026581 on Build GRCh38
rs570958810 Chr.6: 167026530-167026530 on Build GRCh38
rs751564366 Chr.6: 167026498-167026499 on Build GRCh38
rs748210067 Chr.6: 167026572-167026572 on Build GRCh38
rs368498505 Chr.6: 167026520-167026520 on Build GRCh38
rs746735007 Chr.6: 167026604-167026604 on Build GRCh38
rs376639748 Chr.6: 167026645-167026645 on Build GRCh38
rs145310318 Chr.6: 167026549-167026549 on Build GRCh38
rs147558146 Chr.6: 167026576-167026576 on Build GRCh38
rs528556431 Chr.6: 167026563-167026563 on Build GRCh38
rs779295438 Chr.6: 167026644-167026644 on Build GRCh38

View Full Product Details