Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102043906-102044143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC53
Entrez Gene ID
51019
Gene Name
coiled-coil domain containing 53
Gene Aliases
CGI-116
UniGene
Hs.405692
Gene Chromosome Location
Chr.12: 102012840-102062124 on Build GRCh38
Transcripts
NM_001301107.1 NM_016053.3 XM_011538423.1 XM_017019383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
430183 c.324+1G>T p.? Chr.12: 102044104-102044104 on Build GRCh38
6324790 c.306C>A p.A102A Chr.12: 102044123-102044123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778173624 Chr.12: 102044099-102044102 on Build GRCh38
rs532637842 Chr.12: 102044121-102044121 on Build GRCh38
rs370467761 Chr.12: 102044084-102044084 on Build GRCh38
rs552905935 Chr.12: 102043986-102043986 on Build GRCh38
rs147510623 Chr.12: 102044100-102044100 on Build GRCh38
rs773115383 Chr.12: 102043982-102043982 on Build GRCh38
rs765300992 Chr.12: 102044075-102044075 on Build GRCh38
rs756751291 Chr.12: 102044115-102044115 on Build GRCh38
rs760667950 Chr.12: 102044073-102044073 on Build GRCh38
rs777400483 Chr.12: 102044120-102044120 on Build GRCh38
rs566850310 Chr.12: 102044105-102044105 on Build GRCh38
rs775235274 Chr.12: 102044063-102044063 on Build GRCh38
rs748588154 Chr.12: 102044110-102044110 on Build GRCh38
rs73382854 Chr.12: 102044085-102044085 on Build GRCh38
rs750539269 Chr.12: 102044076-102044076 on Build GRCh38
rs1020263749 Chr.12: 102043967-102043967 on Build GRCh38
rs554926968 Chr.12: 102044006-102044007 on Build GRCh38
rs542882642 Chr.12: 102044103-102044103 on Build GRCh38
rs561428463 Chr.12: 102044018-102044018 on Build GRCh38
rs115247989 Chr.12: 102043981-102043981 on Build GRCh38
rs913686586 Chr.12: 102044089-102044089 on Build GRCh38

View Full Product Details