Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40800929-40801160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGLN2
Entrez Gene ID
112398
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
UniGene
Hs.515417
Gene Chromosome Location
Chr.19: 40799143-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Gene Symbol
RAB4B-EGLN2
Entrez Gene ID
100529264
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
Gene Aliases
RERT-lncRNA
UniGene
Hs.631539
Gene Chromosome Location
Chr.19: 40778219-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295989 c.430G>T p.A144S Chr.19: 40801002-40801002 on Build GRCh38
7274073 c.529C>T p.L177L Chr.19: 40801101-40801101 on Build GRCh38
4078377 c.366C>A p.A122A Chr.19: 40800938-40800938 on Build GRCh38
1393895 c.577G>A p.V193I Chr.19: 40801149-40801149 on Build GRCh38
6652306 c.522C>A p.P174P Chr.19: 40801094-40801094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767345570 Chr.19: 40801019-40801019 on Build GRCh38
rs747213479 Chr.19: 40801089-40801089 on Build GRCh38
rs566229682 Chr.19: 40800957-40800957 on Build GRCh38
rs778659160 Chr.19: 40801059-40801059 on Build GRCh38
rs766236137 Chr.19: 40800965-40800965 on Build GRCh38
rs146754458 Chr.19: 40801139-40801139 on Build GRCh38
rs926643907 Chr.19: 40800954-40800954 on Build GRCh38
rs770957227 Chr.19: 40800972-40800972 on Build GRCh38
rs376266825 Chr.19: 40801055-40801055 on Build GRCh38
rs935705560 Chr.19: 40801101-40801101 on Build GRCh38
rs747037763 Chr.19: 40801140-40801140 on Build GRCh38
rs1014086658 Chr.19: 40801124-40801124 on Build GRCh38
rs202088727 Chr.19: 40801020-40801020 on Build GRCh38
rs201258123 Chr.19: 40801051-40801051 on Build GRCh38
rs765288136 Chr.19: 40801131-40801131 on Build GRCh38
rs376790059 Chr.19: 40801090-40801090 on Build GRCh38
rs745886559 Chr.19: 40800983-40800983 on Build GRCh38
rs749225346 Chr.19: 40800970-40800970 on Build GRCh38
rs745968202 Chr.19: 40801068-40801068 on Build GRCh38
rs778285867 Chr.19: 40801023-40801028 on Build GRCh38
rs751089985 Chr.19: 40801008-40801008 on Build GRCh38
rs761099207 Chr.19: 40800999-40800999 on Build GRCh38
rs200595622 Chr.19: 40801091-40801091 on Build GRCh38
rs747637096 Chr.19: 40800955-40800955 on Build GRCh38
rs867933915 Chr.19: 40801018-40801018 on Build GRCh38
rs780315358 Chr.19: 40801079-40801079 on Build GRCh38
rs777041827 Chr.19: 40801003-40801003 on Build GRCh38
rs371954705 Chr.19: 40801060-40801060 on Build GRCh38
rs1052892437 Chr.19: 40801107-40801107 on Build GRCh38
rs551623860 Chr.19: 40801094-40801094 on Build GRCh38
rs775321804 Chr.19: 40801100-40801100 on Build GRCh38
rs763604374 Chr.19: 40801114-40801114 on Build GRCh38
rs369347048 Chr.19: 40801099-40801099 on Build GRCh38
rs117916638 Chr.19: 40801119-40801119 on Build GRCh38
rs777913324 Chr.19: 40800969-40800969 on Build GRCh38
rs759048732 Chr.19: 40801009-40801009 on Build GRCh38
rs759173256 Chr.19: 40801097-40801097 on Build GRCh38
rs750349808 Chr.19: 40801132-40801132 on Build GRCh38
rs768997902 Chr.19: 40801000-40801000 on Build GRCh38
rs898811701 Chr.19: 40801083-40801083 on Build GRCh38
rs760435981 Chr.19: 40801106-40801106 on Build GRCh38
rs753538171 Chr.19: 40801046-40801046 on Build GRCh38
rs376569262 Chr.19: 40801064-40801064 on Build GRCh38
rs139802237 Chr.19: 40801078-40801078 on Build GRCh38
rs756020588 Chr.19: 40801024-40801024 on Build GRCh38
rs561741910 Chr.19: 40800989-40800989 on Build GRCh38
rs779083045 Chr.19: 40800981-40800981 on Build GRCh38
rs139096559 Chr.19: 40800987-40800987 on Build GRCh38
rs373843220 Chr.19: 40801118-40801118 on Build GRCh38
rs758526793 Chr.19: 40801138-40801138 on Build GRCh38
rs773586574 Chr.19: 40801095-40801095 on Build GRCh38
rs144903698 Chr.19: 40801027-40801027 on Build GRCh38
rs1034071713 Chr.19: 40801026-40801026 on Build GRCh38
rs922857701 Chr.19: 40800995-40800995 on Build GRCh38

View Full Product Details