Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32169922-32170157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1 NR_106779.1

Gene Symbol
MIR6721
Entrez Gene ID
102466190
Gene Name
microRNA 6721
Gene Aliases
hsa-mir-6721, mir-6721
UniGene
Gene Chromosome Location
Chr.6: 32170030-32170116 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1 NR_106779.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047511 c.628A>G p.M210V Chr.6: 32170017-32170017 on Build GRCh38
3625340 c.633C>T p.S211S Chr.6: 32170012-32170012 on Build GRCh38
6596356 c.646T>G p.F216V Chr.6: 32169999-32169999 on Build GRCh38
6364017 c.619C>T p.P207S Chr.6: 32170026-32170026 on Build GRCh38
5687083 c.655A>C p.K219Q Chr.6: 32169990-32169990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768730436 Chr.6: 32170135-32170135 on Build GRCh38
rs756987321 Chr.6: 32170099-32170099 on Build GRCh38
rs745821083 Chr.6: 32170126-32170126 on Build GRCh38
rs773164497 Chr.6: 32169980-32169980 on Build GRCh38
rs778319903 Chr.6: 32170075-32170075 on Build GRCh38
rs1008655790 Chr.6: 32170033-32170033 on Build GRCh38
rs375824577 Chr.6: 32169959-32169959 on Build GRCh38
rs771955233 Chr.6: 32169975-32169975 on Build GRCh38
rs149893502 Chr.6: 32169971-32169971 on Build GRCh38
rs758101925 Chr.6: 32170091-32170091 on Build GRCh38
rs754446230 Chr.6: 32170071-32170071 on Build GRCh38
rs147585779 Chr.6: 32169986-32169986 on Build GRCh38
rs997479168 Chr.6: 32170115-32170115 on Build GRCh38
rs777608790 Chr.6: 32170096-32170096 on Build GRCh38
rs763029554 Chr.6: 32170017-32170017 on Build GRCh38
rs777515321 Chr.6: 32169951-32169951 on Build GRCh38
rs776489092 Chr.6: 32170015-32170015 on Build GRCh38
rs778572165 Chr.6: 32169960-32169960 on Build GRCh38
rs1027228711 Chr.6: 32170128-32170128 on Build GRCh38
rs143961469 Chr.6: 32170030-32170030 on Build GRCh38
rs767772319 Chr.6: 32170032-32170032 on Build GRCh38
rs558682365 Chr.6: 32170012-32170012 on Build GRCh38
rs11553431 Chr.6: 32170024-32170024 on Build GRCh38
rs781116036 Chr.6: 32170123-32170123 on Build GRCh38
rs750751399 Chr.6: 32170048-32170048 on Build GRCh38
rs770861940 Chr.6: 32170103-32170103 on Build GRCh38
rs747789210 Chr.6: 32170090-32170090 on Build GRCh38
rs900015643 Chr.6: 32170107-32170107 on Build GRCh38
rs756542128 Chr.6: 32170055-32170055 on Build GRCh38
rs749150815 Chr.6: 32170059-32170059 on Build GRCh38
rs754317834 Chr.6: 32170068-32170068 on Build GRCh38
rs747761876 Chr.6: 32169967-32169967 on Build GRCh38
rs530048326 Chr.6: 32170119-32170119 on Build GRCh38
rs187833281 Chr.6: 32170102-32170102 on Build GRCh38

View Full Product Details