Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60689575-60689848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853138 c.214G>A p.D72N Chr.17: 60689761-60689761 on Build GRCh38
3717503 c.146G>A p.S49N Chr.17: 60689693-60689693 on Build GRCh38
7308900 c.196G>T p.E66* Chr.17: 60689743-60689743 on Build GRCh38
5853139 c.214G>A p.D72N Chr.17: 60689761-60689761 on Build GRCh38
1725307 c.196G>T p.E66* Chr.17: 60689743-60689743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780697275 Chr.17: 60689709-60689709 on Build GRCh38
rs1054195862 Chr.17: 60689620-60689620 on Build GRCh38
rs575831594 Chr.17: 60689755-60689755 on Build GRCh38
rs368323517 Chr.17: 60689786-60689786 on Build GRCh38
rs762041579 Chr.17: 60689804-60689804 on Build GRCh38
rs763995597 Chr.17: 60689798-60689801 on Build GRCh38
rs369736992 Chr.17: 60689667-60689667 on Build GRCh38
rs774977956 Chr.17: 60689635-60689635 on Build GRCh38
rs1026941572 Chr.17: 60689798-60689798 on Build GRCh38
rs936774964 Chr.17: 60689617-60689617 on Build GRCh38
rs372481294 Chr.17: 60689787-60689787 on Build GRCh38
rs767593694 Chr.17: 60689658-60689658 on Build GRCh38
rs771374863 Chr.17: 60689758-60689758 on Build GRCh38
rs1042602924 Chr.17: 60689601-60689601 on Build GRCh38
rs374398691 Chr.17: 60689775-60689775 on Build GRCh38
rs746320029 Chr.17: 60689770-60689770 on Build GRCh38
rs774609545 Chr.17: 60689767-60689767 on Build GRCh38
rs1042076374 Chr.17: 60689643-60689643 on Build GRCh38
rs781380210 Chr.17: 60689696-60689696 on Build GRCh38
rs751478102 Chr.17: 60689694-60689694 on Build GRCh38
rs375041824 Chr.17: 60689680-60689680 on Build GRCh38
rs547781377 Chr.17: 60689800-60689800 on Build GRCh38
rs755754115 Chr.17: 60689725-60689725 on Build GRCh38
rs759982737 Chr.17: 60689656-60689656 on Build GRCh38
rs924009593 Chr.17: 60689605-60689605 on Build GRCh38
rs752762362 Chr.17: 60689673-60689674 on Build GRCh38
rs754879954 Chr.17: 60689699-60689699 on Build GRCh38
rs764284417 Chr.17: 60689676-60689676 on Build GRCh38
rs772469029 Chr.17: 60689781-60689781 on Build GRCh38
rs779869339 Chr.17: 60689691-60689691 on Build GRCh38
rs777539084 Chr.17: 60689736-60689736 on Build GRCh38
rs372774453 Chr.17: 60689716-60689716 on Build GRCh38
rs558259950 Chr.17: 60689627-60689627 on Build GRCh38
rs752722438 Chr.17: 60689664-60689664 on Build GRCh38
rs912347312 Chr.17: 60689615-60689615 on Build GRCh38
rs748999070 Chr.17: 60689748-60689748 on Build GRCh38

View Full Product Details