Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100469273-100469534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS3
Entrez Gene ID
204219
Gene Name
ceramide synthase 3
Gene Aliases
ARCI9, LASS3
UniGene
Hs.662371
Gene Chromosome Location
Chr.15: 100400395-100544982 on Build GRCh38
Transcripts
NM_001290341.2 NM_001290342.2 NM_001290343.1 NM_178842.4 XM_011521355.2 XM_011521357.2 XM_011521358.1 XM_017022002.1 XM_017022003.1 XM_017022004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627355 c.808A>C p.I270L Chr.15: 100469415-100469415 on Build GRCh38
5892250 c.785C>T p.T262I Chr.15: 100469438-100469438 on Build GRCh38
4402203 c.833T>G p.V278G Chr.15: 100469390-100469390 on Build GRCh38
137633 c.838C>T p.P280S Chr.15: 100469385-100469385 on Build GRCh38
6849313 c.799T>C p.F267L Chr.15: 100469424-100469424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015009742 Chr.15: 100469447-100469447 on Build GRCh38
rs867892206 Chr.15: 100469385-100469385 on Build GRCh38
rs768271613 Chr.15: 100469395-100469395 on Build GRCh38
rs74041452 Chr.15: 100469367-100469367 on Build GRCh38
rs760569562 Chr.15: 100469499-100469499 on Build GRCh38
rs747356174 Chr.15: 100469366-100469366 on Build GRCh38
rs761082211 Chr.15: 100469326-100469326 on Build GRCh38
rs114499429 Chr.15: 100469415-100469415 on Build GRCh38
rs778468756 Chr.15: 100469479-100469479 on Build GRCh38
rs781730763 Chr.15: 100469444-100469444 on Build GRCh38
rs779586622 Chr.15: 100469459-100469459 on Build GRCh38
rs755487846 Chr.15: 100469347-100469347 on Build GRCh38
rs752795618 Chr.15: 100469421-100469421 on Build GRCh38
rs374685247 Chr.15: 100469464-100469464 on Build GRCh38
rs557826222 Chr.15: 100469390-100469390 on Build GRCh38
rs754016611 Chr.15: 100469334-100469334 on Build GRCh38
rs745390684 Chr.15: 100469380-100469380 on Build GRCh38
rs771715153 Chr.15: 100469482-100469482 on Build GRCh38
rs772962880 Chr.15: 100469416-100469416 on Build GRCh38
rs920821835 Chr.15: 100469489-100469489 on Build GRCh38
rs764401429 Chr.15: 100469437-100469437 on Build GRCh38
rs758867607 Chr.15: 100469341-100469341 on Build GRCh38
rs200925198 Chr.15: 100469438-100469438 on Build GRCh38
rs758743159 Chr.15: 100469426-100469426 on Build GRCh38
rs568564529 Chr.15: 100469316-100469316 on Build GRCh38
rs12909732 Chr.15: 100469325-100469325 on Build GRCh38
rs76863389 Chr.15: 100469338-100469338 on Build GRCh38
rs757498933 Chr.15: 100469439-100469439 on Build GRCh38
rs769131621 Chr.15: 100469382-100469382 on Build GRCh38
rs1010076284 Chr.15: 100469321-100469321 on Build GRCh38
rs368872348 Chr.15: 100469505-100469505 on Build GRCh38
rs765193339 Chr.15: 100469420-100469420 on Build GRCh38
rs143625266 Chr.15: 100469399-100469399 on Build GRCh38
rs747949926 Chr.15: 100469480-100469480 on Build GRCh38
rs1042961372 Chr.15: 100469324-100469324 on Build GRCh38
rs777328609 Chr.15: 100469418-100469418 on Build GRCh38
rs761591750 Chr.15: 100469400-100469400 on Build GRCh38
rs750852537 Chr.15: 100469452-100469452 on Build GRCh38
rs778322939 Chr.15: 100469359-100469359 on Build GRCh38
rs768363478 Chr.15: 100469467-100469467 on Build GRCh38
rs77590344 Chr.15: 100469488-100469488 on Build GRCh38
rs773233152 Chr.15: 100469498-100469498 on Build GRCh38
rs112307246 Chr.15: 100469319-100469319 on Build GRCh38
rs146792407 Chr.15: 100469419-100469419 on Build GRCh38
rs756608778 Chr.15: 100469453-100469453 on Build GRCh38

View Full Product Details