Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37737844-37738113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
UniGene
Hs.705490
Gene Chromosome Location
Chr.8: 37736579-37757801 on Build GRCh38
Transcripts
NM_001003790.3 NM_001003791.2 NM_007175.6 XM_005273392.2 XM_006716280.2 XM_017013000.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6516086 c.107G>A p.R36K Chr.8: 37738029-37738029 on Build GRCh38
4611289 c.52_53delTC p.F20fs*40 Chr.8: 37737974-37737975 on Build GRCh38
5822454 c.35C>T p.S12F Chr.8: 37737957-37737957 on Build GRCh38
7182465 c.85_88delGGAC p.G29fs*12 Chr.8: 37738007-37738010 on Build GRCh38
4611288 c.52_53delTC p.F20fs*40 Chr.8: 37737974-37737975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986094392 Chr.8: 37737948-37737948 on Build GRCh38
rs965133805 Chr.8: 37737940-37737940 on Build GRCh38
rs886041612 Chr.8: 37737939-37737939 on Build GRCh38
rs745499650 Chr.8: 37738058-37738058 on Build GRCh38
rs770503265 Chr.8: 37737937-37737937 on Build GRCh38
rs976467673 Chr.8: 37737992-37737992 on Build GRCh38
rs754858099 Chr.8: 37737989-37737989 on Build GRCh38
rs760464689 Chr.8: 37737866-37737866 on Build GRCh38
rs772582988 Chr.8: 37737921-37737921 on Build GRCh38
rs746810438 Chr.8: 37737875-37737875 on Build GRCh38
rs748772104 Chr.8: 37738053-37738053 on Build GRCh38
rs748766759 Chr.8: 37737930-37737930 on Build GRCh38
rs1028469104 Chr.8: 37737995-37737995 on Build GRCh38
rs781111255 Chr.8: 37738017-37738017 on Build GRCh38
rs773945506 Chr.8: 37737944-37737944 on Build GRCh38
rs772488282 Chr.8: 37738070-37738070 on Build GRCh38
rs1020709705 Chr.8: 37737906-37737906 on Build GRCh38
rs781307502 Chr.8: 37737889-37737889 on Build GRCh38
rs754767980 Chr.8: 37737878-37737878 on Build GRCh38
rs369927330 Chr.8: 37738043-37738044 on Build GRCh38
rs376645631 Chr.8: 37738056-37738056 on Build GRCh38
rs764171662 Chr.8: 37737954-37737955 on Build GRCh38
rs765953850 Chr.8: 37737916-37737916 on Build GRCh38
rs776029831 Chr.8: 37737957-37737957 on Build GRCh38
rs368986092 Chr.8: 37737961-37737961 on Build GRCh38
rs938835638 Chr.8: 37738083-37738083 on Build GRCh38
rs570095242 Chr.8: 37737977-37737977 on Build GRCh38
rs77176932 Chr.8: 37737951-37737951 on Build GRCh38
rs371427544 Chr.8: 37738030-37738030 on Build GRCh38
rs756264790 Chr.8: 37737870-37737870 on Build GRCh38
rs368897065 Chr.8: 37737968-37737968 on Build GRCh38
rs530067147 Chr.8: 37737871-37737871 on Build GRCh38
rs886042197 Chr.8: 37737958-37737958 on Build GRCh38
rs770390076 Chr.8: 37738054-37738054 on Build GRCh38
rs748173991 Chr.8: 37738018-37738018 on Build GRCh38
rs374800981 Chr.8: 37737949-37737950 on Build GRCh38
rs548413025 Chr.8: 37737893-37737893 on Build GRCh38
rs765575293 Chr.8: 37737969-37737969 on Build GRCh38
rs761077621 Chr.8: 37737959-37737959 on Build GRCh38
rs777708321 Chr.8: 37738046-37738046 on Build GRCh38
rs753863065 Chr.8: 37737963-37737963 on Build GRCh38
rs200790467 Chr.8: 37737954-37737954 on Build GRCh38

View Full Product Details