Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 830273-830520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2B
Entrez Gene ID
283229
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
UniGene
Hs.660936
Gene Chromosome Location
Chr.11: 825558-831991 on Build GRCh38
Transcripts
NM_001286606.1 NM_173584.4 XM_011520011.1 XM_011520019.1 XM_017017581.1 XM_017017582.1 XM_017017583.1 XM_017017584.1 XM_017017585.1 XM_017017586.1 XM_017017587.1 XM_017017588.1 XM_017017589.1 XM_017017590.1 XM_017017591.1 XM_017017592.1 XM_017017593.1 XM_017017594.1 XM_017017595.1 XM_017017596.1 XM_017017597.1 XM_017017598.1 XM_017017599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1357339 c.635G>A p.R212H Chr.11: 830279-830279 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762372849 Chr.11: 830469-830482 on Build GRCh38
rs745321905 Chr.11: 830294-830294 on Build GRCh38
rs974992018 Chr.11: 830429-830429 on Build GRCh38
rs907614742 Chr.11: 830478-830478 on Build GRCh38
rs761010299 Chr.11: 830305-830305 on Build GRCh38
rs769188779 Chr.11: 830297-830297 on Build GRCh38
rs779515440 Chr.11: 830470-830470 on Build GRCh38
rs72847210 Chr.11: 830383-830383 on Build GRCh38
rs766853619 Chr.11: 830306-830306 on Build GRCh38
rs1018916380 Chr.11: 830422-830422 on Build GRCh38
rs964863946 Chr.11: 830410-830410 on Build GRCh38
rs113266690 Chr.11: 830497-830497 on Build GRCh38
rs533498453 Chr.11: 830464-830464 on Build GRCh38
rs996670603 Chr.11: 830457-830457 on Build GRCh38
rs781114304 Chr.11: 830424-830424 on Build GRCh38
rs4075290 Chr.11: 830487-830487 on Build GRCh38
rs773534255 Chr.11: 830304-830304 on Build GRCh38
rs745819089 Chr.11: 830444-830444 on Build GRCh38
rs1031714351 Chr.11: 830330-830330 on Build GRCh38
rs759622340 Chr.11: 830321-830321 on Build GRCh38
rs983557955 Chr.11: 830473-830473 on Build GRCh38
rs765430831 Chr.11: 830363-830363 on Build GRCh38
rs529330631 Chr.11: 830476-830476 on Build GRCh38
rs916390084 Chr.11: 830499-830499 on Build GRCh38
rs752877413 Chr.11: 830377-830377 on Build GRCh38
rs867577133 Chr.11: 830373-830373 on Build GRCh38
rs1015213313 Chr.11: 830495-830495 on Build GRCh38
rs758667190 Chr.11: 830380-830380 on Build GRCh38
rs1003288336 Chr.11: 830309-830309 on Build GRCh38
rs1011686598 Chr.11: 830388-830388 on Build GRCh38
rs188032561 Chr.11: 830298-830298 on Build GRCh38
rs550106508 Chr.11: 830399-830399 on Build GRCh38
rs768345389 Chr.11: 830299-830299 on Build GRCh38
rs893639112 Chr.11: 830331-830332 on Build GRCh38
rs777009500 Chr.11: 830313-830313 on Build GRCh38
rs139814220 Chr.11: 830465-830465 on Build GRCh38
rs757013223 Chr.11: 830409-830409 on Build GRCh38
rs541774564 Chr.11: 830290-830290 on Build GRCh38
rs947890172 Chr.11: 830500-830500 on Build GRCh38

View Full Product Details