Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18412603-18412842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZANK1
Entrez Gene ID
55184
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
UniGene
Hs.472225
Gene Chromosome Location
Chr.20: 18383367-18467185 on Build GRCh38
Transcripts
NM_001099407.1 XM_005260742.3 XM_005260743.3 XM_006723584.2 XM_011529268.2 XM_011529271.2 XM_011529272.2 XM_011529275.2 XM_011529277.2 XM_017027909.1 XM_017027910.1 XM_017027911.1 XM_017027912.1 XM_017027913.1 XM_017027914.1 XM_017027915.1 XM_017027916.1 XM_017027917.1 XM_017027918.1 XM_017027919.1 XM_017027920.1 XM_017027921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286958 c.1013A>G p.E338G Chr.20: 18412723-18412723 on Build GRCh38
6383248 c.913G>A p.A305T Chr.20: 18412664-18412664 on Build GRCh38
6304206 c.1037A>G p.Q346R Chr.20: 18412699-18412699 on Build GRCh38
6368104 c.839C>A p.A280E Chr.20: 18412738-18412738 on Build GRCh38
2704544 c.1346delA p.K449fs*5 Chr.20: 18412732-18412732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374735349 Chr.20: 18412747-18412747 on Build GRCh38
rs747295212 Chr.20: 18412761-18412761 on Build GRCh38
rs886593550 Chr.20: 18412677-18412677 on Build GRCh38
rs6075337 Chr.20: 18412668-18412668 on Build GRCh38
rs374751305 Chr.20: 18412666-18412666 on Build GRCh38
rs1006501186 Chr.20: 18412705-18412705 on Build GRCh38
rs1045439508 Chr.20: 18412688-18412688 on Build GRCh38
rs59695827 Chr.20: 18412631-18412631 on Build GRCh38
rs772263567 Chr.20: 18412633-18412633 on Build GRCh38
rs763489712 Chr.20: 18412803-18412803 on Build GRCh38
rs754779331 Chr.20: 18412750-18412750 on Build GRCh38
rs761421436 Chr.20: 18412813-18412813 on Build GRCh38
rs577379587 Chr.20: 18412756-18412756 on Build GRCh38
rs774845008 Chr.20: 18412812-18412812 on Build GRCh38
rs755069971 Chr.20: 18412670-18412670 on Build GRCh38
rs34085671 Chr.20: 18412627-18412628 on Build GRCh38
rs769211912 Chr.20: 18412806-18412806 on Build GRCh38
rs745994799 Chr.20: 18412781-18412781 on Build GRCh38
rs758915443 Chr.20: 18412783-18412783 on Build GRCh38
rs542831269 Chr.20: 18412671-18412671 on Build GRCh38
rs778923173 Chr.20: 18412731-18412732 on Build GRCh38
rs756066338 Chr.20: 18412629-18412629 on Build GRCh38
rs771167934 Chr.20: 18412773-18412773 on Build GRCh38
rs996587292 Chr.20: 18412784-18412784 on Build GRCh38
rs749927645 Chr.20: 18412722-18412722 on Build GRCh38
rs770918012 Chr.20: 18412659-18412659 on Build GRCh38
rs557101505 Chr.20: 18412741-18412741 on Build GRCh38
rs748374248 Chr.20: 18412795-18412796 on Build GRCh38
rs778609869 Chr.20: 18412754-18412754 on Build GRCh38
rs749051406 Chr.20: 18412637-18412637 on Build GRCh38
rs900725260 Chr.20: 18412749-18412749 on Build GRCh38
rs865859131 Chr.20: 18412775-18412775 on Build GRCh38
rs763436655 Chr.20: 18412689-18412689 on Build GRCh38
rs776104671 Chr.20: 18412683-18412683 on Build GRCh38
rs766995132 Chr.20: 18412720-18412720 on Build GRCh38
rs756046356 Chr.20: 18412742-18412742 on Build GRCh38
rs776157744 Chr.20: 18412789-18412789 on Build GRCh38
rs906928775 Chr.20: 18412731-18412732 on Build GRCh38
rs764497661 Chr.20: 18412706-18412706 on Build GRCh38
rs768398583 Chr.20: 18412639-18412639 on Build GRCh38
rs747174497 Chr.20: 18412651-18412651 on Build GRCh38
rs777991861 Chr.20: 18412645-18412645 on Build GRCh38
rs761524785 Chr.20: 18412716-18412716 on Build GRCh38
rs769717059 Chr.20: 18412786-18412786 on Build GRCh38
rs781173651 Chr.20: 18412780-18412780 on Build GRCh38
rs751264370 Chr.20: 18412712-18412712 on Build GRCh38
rs754309818 Chr.20: 18412647-18412648 on Build GRCh38

View Full Product Details