Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69338504-69338763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT16
Entrez Gene ID
57452
Gene Name
polypeptide N-acetylgalactosaminyltransferase 16
Gene Aliases
GALNACT16, GALNTL1, GalNAc-T16
UniGene
Hs.21035
Gene Chromosome Location
Chr.14: 69259546-69379904 on Build GRCh38
Transcripts
NM_001168368.1 NM_020692.2 XM_011537005.1 XM_011537006.2 XM_011537007.2 XM_017021498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6216989 c.1014C>T p.V338V Chr.14: 69338697-69338697 on Build GRCh38
3497371 c.1011C>T p.I337I Chr.14: 69338694-69338694 on Build GRCh38
7369172 c.1049G>A p.R350Q Chr.14: 69338732-69338732 on Build GRCh38
4052028 c.997G>T p.G333C Chr.14: 69338680-69338680 on Build GRCh38
5954588 c.995G>A p.G332D Chr.14: 69338678-69338678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145531208 Chr.14: 69338578-69338578 on Build GRCh38
rs910105177 Chr.14: 69338546-69338546 on Build GRCh38
rs769773991 Chr.14: 69338633-69338633 on Build GRCh38
rs755771930 Chr.14: 69338685-69338685 on Build GRCh38
rs201839131 Chr.14: 69338646-69338646 on Build GRCh38
rs926664285 Chr.14: 69338532-69338532 on Build GRCh38
rs146506053 Chr.14: 69338665-69338665 on Build GRCh38
rs768663748 Chr.14: 69338667-69338667 on Build GRCh38
rs762084731 Chr.14: 69338678-69338678 on Build GRCh38
rs767036232 Chr.14: 69338549-69338549 on Build GRCh38
rs546569575 Chr.14: 69338552-69338552 on Build GRCh38
rs373202693 Chr.14: 69338632-69338632 on Build GRCh38
rs200145110 Chr.14: 69338672-69338672 on Build GRCh38
rs556954532 Chr.14: 69338694-69338694 on Build GRCh38
rs1021510451 Chr.14: 69338707-69338707 on Build GRCh38
rs781191279 Chr.14: 69338627-69338627 on Build GRCh38
rs764174616 Chr.14: 69338671-69338671 on Build GRCh38
rs766935197 Chr.14: 69338697-69338697 on Build GRCh38
rs567508249 Chr.14: 69338529-69338529 on Build GRCh38
rs201006835 Chr.14: 69338689-69338689 on Build GRCh38
rs376166666 Chr.14: 69338731-69338731 on Build GRCh38
rs757114530 Chr.14: 69338619-69338619 on Build GRCh38
rs187385915 Chr.14: 69338539-69338539 on Build GRCh38
rs945607679 Chr.14: 69338617-69338617 on Build GRCh38
rs984389359 Chr.14: 69338541-69338541 on Build GRCh38
rs904007324 Chr.14: 69338639-69338639 on Build GRCh38
rs375325329 Chr.14: 69338640-69338640 on Build GRCh38
rs751652984 Chr.14: 69338635-69338637 on Build GRCh38
rs774598680 Chr.14: 69338674-69338674 on Build GRCh38
rs75891218 Chr.14: 69338695-69338695 on Build GRCh38
rs756047816 Chr.14: 69338638-69338639 on Build GRCh38
rs370547085 Chr.14: 69338732-69338732 on Build GRCh38
rs538719612 Chr.14: 69338589-69338589 on Build GRCh38
rs867505008 Chr.14: 69338724-69338724 on Build GRCh38
rs750113776 Chr.14: 69338648-69338648 on Build GRCh38
rs772368943 Chr.14: 69338684-69338684 on Build GRCh38
rs754354389 Chr.14: 69338700-69338700 on Build GRCh38
rs751244158 Chr.14: 69338743-69338743 on Build GRCh38

View Full Product Details