Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46182473-46182746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATC1L
Entrez Gene ID
84221
Gene Name
spermatogenesis and centriole associated 1-like
Gene Aliases
C21orf56
UniGene
Hs.381214
Gene Chromosome Location
Chr.21: 46161148-46185159 on Build GRCh38
Transcripts
NM_001142854.1 NM_032261.4 XM_005261188.4 XM_017028480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6038520 c.155C>T p.A52V Chr.21: 46182662-46182662 on Build GRCh38
4989012 c.135C>G p.H45Q Chr.21: 46182682-46182682 on Build GRCh38
5565666 c.97C>T p.R33W Chr.21: 46182720-46182720 on Build GRCh38
4797176 c.186G>A p.G62G Chr.21: 46182631-46182631 on Build GRCh38
7301718 c.126C>T p.G42G Chr.21: 46182691-46182691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535746289 Chr.21: 46182643-46182643 on Build GRCh38
rs1001961658 Chr.21: 46182633-46182633 on Build GRCh38
rs973736433 Chr.21: 46182688-46182688 on Build GRCh38
rs878961892 Chr.21: 46182582-46182582 on Build GRCh38
rs766841416 Chr.21: 46182628-46182629 on Build GRCh38
rs774467955 Chr.21: 46182618-46182618 on Build GRCh38
rs900648713 Chr.21: 46182614-46182614 on Build GRCh38
rs761333673 Chr.21: 46182634-46182634 on Build GRCh38
rs138704000 Chr.21: 46182561-46182561 on Build GRCh38
rs780631215 Chr.21: 46182720-46182720 on Build GRCh38
rs745365276 Chr.21: 46182722-46182722 on Build GRCh38
rs983449018 Chr.21: 46182723-46182723 on Build GRCh38
rs779300435 Chr.21: 46182690-46182690 on Build GRCh38
rs543964172 Chr.21: 46182579-46182579 on Build GRCh38
rs755367806 Chr.21: 46182667-46182667 on Build GRCh38
rs1033489758 Chr.21: 46182638-46182638 on Build GRCh38
rs537523590 Chr.21: 46182622-46182622 on Build GRCh38
rs942110944 Chr.21: 46182580-46182580 on Build GRCh38
rs759406154 Chr.21: 46182708-46182708 on Build GRCh38
rs751196576 Chr.21: 46182692-46182692 on Build GRCh38
rs973808746 Chr.21: 46182549-46182549 on Build GRCh38
rs527851540 Chr.21: 46182529-46182529 on Build GRCh38
rs750444314 Chr.21: 46182687-46182687 on Build GRCh38
rs1013488971 Chr.21: 46182659-46182659 on Build GRCh38
rs762711263 Chr.21: 46182613-46182613 on Build GRCh38
rs570245905 Chr.21: 46182573-46182573 on Build GRCh38
rs75252048 Chr.21: 46182518-46182518 on Build GRCh38
rs929330003 Chr.21: 46182555-46182555 on Build GRCh38
rs1052037931 Chr.21: 46182624-46182624 on Build GRCh38
rs908182095 Chr.21: 46182530-46182530 on Build GRCh38
rs939709081 Chr.21: 46182535-46182535 on Build GRCh38
rs960667717 Chr.21: 46182500-46182500 on Build GRCh38
rs768906011 Chr.21: 46182617-46182617 on Build GRCh38
rs761452906 Chr.21: 46182616-46182616 on Build GRCh38
rs553756950 Chr.21: 46182719-46182719 on Build GRCh38
rs754303901 Chr.21: 46182661-46182661 on Build GRCh38
rs891783121 Chr.21: 46182620-46182620 on Build GRCh38
rs917798061 Chr.21: 46182552-46182552 on Build GRCh38
rs368890284 Chr.21: 46182631-46182631 on Build GRCh38
rs773844866 Chr.21: 46182630-46182630 on Build GRCh38
rs549793422 Chr.21: 46182627-46182627 on Build GRCh38
rs912790937 Chr.21: 46182596-46182596 on Build GRCh38
rs762647619 Chr.21: 46182567-46182567 on Build GRCh38
rs992071175 Chr.21: 46182548-46182548 on Build GRCh38

View Full Product Details