Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56051936-56052171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450583 c.71_100del30 p.S25_L34del10 Chr.7: 56051919-56051948 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3735665 Chr.7: 56052037-56052037 on Build GRCh38
rs1005128371 Chr.7: 56052015-56052015 on Build GRCh38
rs551578250 Chr.7: 56052129-56052129 on Build GRCh38
rs764727881 Chr.7: 56052012-56052012 on Build GRCh38
rs993275117 Chr.7: 56052075-56052075 on Build GRCh38
rs997879990 Chr.7: 56051977-56051977 on Build GRCh38
rs899555698 Chr.7: 56051968-56051968 on Build GRCh38
rs867489064 Chr.7: 56052147-56052147 on Build GRCh38
rs767545564 Chr.7: 56051971-56051971 on Build GRCh38
rs1034423890 Chr.7: 56052054-56052054 on Build GRCh38
rs1015559536 Chr.7: 56052017-56052017 on Build GRCh38
rs376612120 Chr.7: 56052035-56052035 on Build GRCh38
rs566851687 Chr.7: 56052106-56052106 on Build GRCh38
rs754137745 Chr.7: 56052021-56052021 on Build GRCh38
rs10243685 Chr.7: 56052028-56052028 on Build GRCh38
rs750960179 Chr.7: 56052149-56052149 on Build GRCh38
rs757686032 Chr.7: 56052023-56052023 on Build GRCh38
rs7780755 Chr.7: 56052143-56052143 on Build GRCh38
rs761299523 Chr.7: 56052132-56052132 on Build GRCh38
rs958761884 Chr.7: 56052063-56052069 on Build GRCh38
rs143477439 Chr.7: 56052004-56052004 on Build GRCh38
rs368306614 Chr.7: 56051992-56051992 on Build GRCh38
rs373223984 Chr.7: 56052061-56052061 on Build GRCh38

View Full Product Details