Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49394415-49394668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC155
Entrez Gene ID
147872
Gene Name
coiled-coil domain containing 155
Gene Aliases
KASH5
UniGene
Hs.662085
Gene Chromosome Location
Chr.19: 49388218-49418002 on Build GRCh38
Transcripts
NM_144688.4 XM_005258549.3 XM_011526488.2 XM_011526489.2 XM_011526490.2 XM_011526494.2 XM_011526495.2 XM_011526496.2 XM_011526497.2 XM_017026333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359825 c.86C>A p.P29Q Chr.19: 49394518-49394518 on Build GRCh38
6621752 c.48C>T p.Y16Y Chr.19: 49394480-49394480 on Build GRCh38
6621751 c.119C>T p.T40M Chr.19: 49394551-49394551 on Build GRCh38
475022 c.58C>T p.R20W Chr.19: 49394490-49394490 on Build GRCh38
6278583 c.69G>A p.E23E Chr.19: 49394501-49394501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867808863 Chr.19: 49394517-49394517 on Build GRCh38
rs79229997 Chr.19: 49394552-49394552 on Build GRCh38
rs759578570 Chr.19: 49394545-49394545 on Build GRCh38
rs866743988 Chr.19: 49394581-49394582 on Build GRCh38
rs779834088 Chr.19: 49394519-49394519 on Build GRCh38
rs983624442 Chr.19: 49394551-49394551 on Build GRCh38
rs199971301 Chr.19: 49394463-49394463 on Build GRCh38
rs773492848 Chr.19: 49394525-49394525 on Build GRCh38
rs201036885 Chr.19: 49394584-49394584 on Build GRCh38
rs778507057 Chr.19: 49394498-49394498 on Build GRCh38
rs758451832 Chr.19: 49394518-49394518 on Build GRCh38
rs749456649 Chr.19: 49394533-49394533 on Build GRCh38
rs761614088 Chr.19: 49394565-49394565 on Build GRCh38
rs769190682 Chr.19: 49394625-49394625 on Build GRCh38
rs558625400 Chr.19: 49394603-49394603 on Build GRCh38
rs751390576 Chr.19: 49394593-49394593 on Build GRCh38
rs769812920 Chr.19: 49394490-49394490 on Build GRCh38
rs368300261 Chr.19: 49394522-49394522 on Build GRCh38
rs777369371 Chr.19: 49394460-49394460 on Build GRCh38
rs374364849 Chr.19: 49394464-49394464 on Build GRCh38
rs753143828 Chr.19: 49394485-49394485 on Build GRCh38
rs764953339 Chr.19: 49394582-49394582 on Build GRCh38
rs376201969 Chr.19: 49394615-49394615 on Build GRCh38
rs373290179 Chr.19: 49394623-49394623 on Build GRCh38
rs753540489 Chr.19: 49394558-49394558 on Build GRCh38
rs547616039 Chr.19: 49394492-49394492 on Build GRCh38
rs774477757 Chr.19: 49394539-49394539 on Build GRCh38
rs367979802 Chr.19: 49394444-49394444 on Build GRCh38
rs200432854 Chr.19: 49394590-49394590 on Build GRCh38
rs756830729 Chr.19: 49394496-49394496 on Build GRCh38
rs367728051 Chr.19: 49394556-49394556 on Build GRCh38
rs1006847719 Chr.19: 49394489-49394489 on Build GRCh38
rs988643974 Chr.19: 49394588-49394588 on Build GRCh38
rs763826408 Chr.19: 49394491-49394491 on Build GRCh38
rs771132124 Chr.19: 49394534-49394534 on Build GRCh38
rs745368612 Chr.19: 49394507-49394507 on Build GRCh38
rs748665033 Chr.19: 49394540-49394544 on Build GRCh38
rs568754480 Chr.19: 49394484-49394484 on Build GRCh38
rs79785583 Chr.19: 49394616-49394616 on Build GRCh38
rs780909379 Chr.19: 49394602-49394602 on Build GRCh38
rs529432324 Chr.19: 49394487-49394487 on Build GRCh38
rs755040277 Chr.19: 49394470-49394470 on Build GRCh38
rs570304611 Chr.19: 49394607-49394607 on Build GRCh38
rs778998926 Chr.19: 49394483-49394484 on Build GRCh38
rs1017286539 Chr.19: 49394549-49394549 on Build GRCh38
rs761132311 Chr.19: 49394555-49394555 on Build GRCh38
rs771179438 Chr.19: 49394606-49394606 on Build GRCh38
rs760097475 Chr.19: 49394476-49394476 on Build GRCh38

View Full Product Details