Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113881806-113882033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Gene Symbol
BCL2L15
Entrez Gene ID
440603
Gene Name
BCL2 like 15
Gene Aliases
Bfk, C1orf178
UniGene
Hs.123106
Gene Chromosome Location
Chr.1: 113876814-113887547 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380371 c.295T>C p.W99R Chr.1: 113881952-113881952 on Build GRCh38
2176788 c.382C>T p.P128S Chr.1: 113881865-113881865 on Build GRCh38
3965711 c.292A>G p.T98A Chr.1: 113881955-113881955 on Build GRCh38
4493252 c.411C>T p.P137P Chr.1: 113881836-113881836 on Build GRCh38
6380370 c.296G>T p.W99L Chr.1: 113881951-113881951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951068160 Chr.1: 113882002-113882002 on Build GRCh38
rs746706671 Chr.1: 113881977-113881977 on Build GRCh38
rs772261515 Chr.1: 113881852-113881864 on Build GRCh38
rs748004126 Chr.1: 113881830-113881830 on Build GRCh38
rs757973338 Chr.1: 113881831-113881831 on Build GRCh38
rs553552734 Chr.1: 113881887-113881887 on Build GRCh38
rs747601726 Chr.1: 113881974-113881974 on Build GRCh38
rs754778320 Chr.1: 113881829-113881829 on Build GRCh38
rs756901135 Chr.1: 113881978-113881978 on Build GRCh38
rs774688441 Chr.1: 113881865-113881865 on Build GRCh38
rs749767273 Chr.1: 113881858-113881858 on Build GRCh38
rs1018138660 Chr.1: 113881922-113881922 on Build GRCh38
rs539072371 Chr.1: 113881944-113881944 on Build GRCh38
rs372905255 Chr.1: 113882005-113882005 on Build GRCh38
rs769150956 Chr.1: 113881992-113881992 on Build GRCh38
rs767565419 Chr.1: 113881871-113881871 on Build GRCh38
rs892495345 Chr.1: 113881856-113881856 on Build GRCh38
rs915961005 Chr.1: 113881872-113881872 on Build GRCh38
rs201714517 Chr.1: 113881927-113881927 on Build GRCh38
rs1217381 Chr.1: 113881979-113881979 on Build GRCh38
rs891053238 Chr.1: 113881895-113881895 on Build GRCh38
rs746446095 Chr.1: 113881841-113881841 on Build GRCh38
rs779068934 Chr.1: 113881828-113881828 on Build GRCh38
rs758227438 Chr.1: 113881954-113881954 on Build GRCh38
rs770499396 Chr.1: 113881848-113881848 on Build GRCh38
rs116085462 Chr.1: 113881940-113881940 on Build GRCh38
rs866403177 Chr.1: 113881999-113881999 on Build GRCh38
rs762309239 Chr.1: 113881869-113881869 on Build GRCh38
rs777487442 Chr.1: 113881839-113881839 on Build GRCh38
rs369636145 Chr.1: 113881988-113881988 on Build GRCh38
rs775937645 Chr.1: 113881857-113881857 on Build GRCh38
rs760620431 Chr.1: 113881906-113881906 on Build GRCh38
rs777242392 Chr.1: 113881968-113881968 on Build GRCh38
rs759581293 Chr.1: 113881884-113881884 on Build GRCh38
rs114941235 Chr.1: 113881987-113881987 on Build GRCh38
rs769202410 Chr.1: 113881864-113881864 on Build GRCh38
rs752535835 Chr.1: 113881945-113881945 on Build GRCh38
rs780394339 Chr.1: 113881962-113881963 on Build GRCh38
rs1007181337 Chr.1: 113881896-113881896 on Build GRCh38
rs745947845 Chr.1: 113881846-113881847 on Build GRCh38

View Full Product Details