Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62627052-62627277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162961 c.2257C>T p.L753L Chr.11: 62627113-62627113 on Build GRCh38
6669218 c.2319G>A p.A773A Chr.11: 62627117-62627117 on Build GRCh38
1676102 c.2290G>T p.G764C Chr.11: 62627080-62627080 on Build GRCh38
6669219 c.2253G>A p.A751A Chr.11: 62627117-62627117 on Build GRCh38
3809848 c.2260G>C p.V754L Chr.11: 62627110-62627110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033238356 Chr.11: 62627167-62627167 on Build GRCh38
rs1014218282 Chr.11: 62627106-62627106 on Build GRCh38
rs757578887 Chr.11: 62627171-62627171 on Build GRCh38
rs771755933 Chr.11: 62627085-62627085 on Build GRCh38
rs1020057727 Chr.11: 62627071-62627071 on Build GRCh38
rs763082802 Chr.11: 62627158-62627158 on Build GRCh38
rs775761544 Chr.11: 62627138-62627138 on Build GRCh38
rs368563939 Chr.11: 62627176-62627176 on Build GRCh38
rs192389541 Chr.11: 62627236-62627236 on Build GRCh38
rs771079554 Chr.11: 62627147-62627148 on Build GRCh38
rs765237116 Chr.11: 62627133-62627133 on Build GRCh38
rs751740452 Chr.11: 62627170-62627170 on Build GRCh38
rs941587240 Chr.11: 62627235-62627235 on Build GRCh38
rs1020214808 Chr.11: 62627107-62627107 on Build GRCh38
rs868816483 Chr.11: 62627121-62627121 on Build GRCh38
rs757863937 Chr.11: 62627257-62627257 on Build GRCh38
rs750746389 Chr.11: 62627175-62627175 on Build GRCh38
rs375923197 Chr.11: 62627187-62627187 on Build GRCh38
rs137897818 Chr.11: 62627193-62627193 on Build GRCh38
rs958575447 Chr.11: 62627162-62627162 on Build GRCh38
rs371022745 Chr.11: 62627246-62627246 on Build GRCh38
rs183266881 Chr.11: 62627243-62627243 on Build GRCh38
rs140219733 Chr.11: 62627118-62627118 on Build GRCh38
rs200456484 Chr.11: 62627117-62627117 on Build GRCh38
rs923427983 Chr.11: 62627203-62627203 on Build GRCh38
rs147361014 Chr.11: 62627182-62627182 on Build GRCh38
rs201994619 Chr.11: 62627093-62627093 on Build GRCh38
rs971828323 Chr.11: 62627197-62627197 on Build GRCh38
rs767980518 Chr.11: 62627174-62627174 on Build GRCh38
rs373852510 Chr.11: 62627255-62627255 on Build GRCh38
rs772872094 Chr.11: 62627086-62627086 on Build GRCh38
rs770691650 Chr.11: 62627099-62627099 on Build GRCh38
rs142739405 Chr.11: 62627180-62627180 on Build GRCh38
rs908807407 Chr.11: 62627201-62627201 on Build GRCh38
rs568832916 Chr.11: 62627194-62627194 on Build GRCh38
rs764446223 Chr.11: 62627169-62627169 on Build GRCh38
rs572016346 Chr.11: 62627248-62627248 on Build GRCh38
rs34969464 Chr.11: 62627177-62627178 on Build GRCh38

View Full Product Details